Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24440935-24441194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADORA2A
Entrez Gene ID
135
Gene Name
adenosine A2a receptor
Gene Aliases
A2aR, ADORA2, RDC8
UniGene
Hs.197029
Gene Chromosome Location
Chr.22: 24423597-24442360 on Build GRCh38
Transcripts
NM_000675.5 NM_001278497.1 NM_001278498.1 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028483.2 NR_028484.2 NR_103546.1

Gene Symbol
ADORA2A-AS1
Entrez Gene ID
646023
Gene Name
ADORA2A antisense RNA 1
Gene Aliases
C22orf45
UniGene
Hs.632761
Gene Chromosome Location
Chr.22: 24429206-24495074 on Build GRCh38
Transcripts
NM_000675.5 NM_001278497.1 NM_001278498.1 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028483.2 NR_028484.2 NR_103546.1

Gene Symbol
SPECC1L-ADORA2A
Entrez Gene ID
101730217
Gene Name
SPECC1L-ADORA2A readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.474384
Gene Chromosome Location
Chr.22: 24270817-24442360 on Build GRCh38
Transcripts
NM_000675.5 NM_001278497.1 NM_001278498.1 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028483.2 NR_028484.2 NR_103546.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3552643 c.885C>T p.F295F Chr.22: 24441135-24441135 on Build GRCh38
5393266 c.826C>T p.L276F Chr.22: 24441076-24441076 on Build GRCh38
3552644 c.938C>T p.P313L Chr.22: 24441188-24441188 on Build GRCh38
6595321 c.877C>T p.R293C Chr.22: 24441127-24441127 on Build GRCh38
4772850 c.777delC p.D261fs*23 Chr.22: 24441027-24441027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043964417 Chr.22: 24440989-24440989 on Build GRCh38
rs779344683 Chr.22: 24441089-24441089 on Build GRCh38
rs767602595 Chr.22: 24441043-24441043 on Build GRCh38
rs200179427 Chr.22: 24441025-24441025 on Build GRCh38
rs777455392 Chr.22: 24441114-24441114 on Build GRCh38
rs200000322 Chr.22: 24441045-24441045 on Build GRCh38
rs371233023 Chr.22: 24440978-24440978 on Build GRCh38
rs201181215 Chr.22: 24441136-24441136 on Build GRCh38
rs999690973 Chr.22: 24441009-24441009 on Build GRCh38
rs141377465 Chr.22: 24441030-24441030 on Build GRCh38
rs4990 Chr.22: 24441149-24441149 on Build GRCh38
rs199666600 Chr.22: 24441161-24441161 on Build GRCh38
rs998572202 Chr.22: 24441153-24441153 on Build GRCh38
rs770667510 Chr.22: 24440985-24440985 on Build GRCh38
rs142560733 Chr.22: 24441127-24441127 on Build GRCh38
rs780201352 Chr.22: 24441068-24441068 on Build GRCh38
rs759348728 Chr.22: 24440996-24440996 on Build GRCh38
rs201077707 Chr.22: 24441115-24441115 on Build GRCh38
rs200323750 Chr.22: 24441084-24441084 on Build GRCh38
rs202222894 Chr.22: 24441119-24441119 on Build GRCh38
rs201680854 Chr.22: 24441033-24441033 on Build GRCh38
rs146268015 Chr.22: 24441031-24441031 on Build GRCh38
rs145970572 Chr.22: 24441129-24441129 on Build GRCh38
rs141942830 Chr.22: 24441073-24441073 on Build GRCh38
rs17650937 Chr.22: 24441135-24441135 on Build GRCh38
rs201173406 Chr.22: 24441072-24441072 on Build GRCh38
rs773303136 Chr.22: 24441132-24441132 on Build GRCh38
rs201025926 Chr.22: 24441128-24441128 on Build GRCh38
rs199792438 Chr.22: 24441137-24441137 on Build GRCh38
rs769621641 Chr.22: 24441122-24441122 on Build GRCh38
rs778398772 Chr.22: 24441167-24441167 on Build GRCh38
rs370669844 Chr.22: 24441123-24441123 on Build GRCh38
rs878978641 Chr.22: 24441166-24441166 on Build GRCh38
rs200799056 Chr.22: 24441168-24441168 on Build GRCh38
rs745714462 Chr.22: 24441121-24441121 on Build GRCh38
rs200117692 Chr.22: 24441169-24441169 on Build GRCh38
rs545585791 Chr.22: 24441093-24441093 on Build GRCh38
rs55801059 Chr.22: 24440991-24440991 on Build GRCh38
rs761858787 Chr.22: 24441034-24441034 on Build GRCh38
rs760407585 Chr.22: 24440975-24440975 on Build GRCh38
rs781051901 Chr.22: 24441173-24441173 on Build GRCh38
rs968793808 Chr.22: 24441147-24441147 on Build GRCh38
rs80266635 Chr.22: 24440961-24440961 on Build GRCh38
rs756360364 Chr.22: 24441059-24441059 on Build GRCh38
rs200512185 Chr.22: 24441120-24441120 on Build GRCh38
rs201666195 Chr.22: 24441160-24441160 on Build GRCh38
rs748699595 Chr.22: 24441092-24441092 on Build GRCh38

View Full Product Details