Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 61820751-61820990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH8
Entrez Gene ID
1006
Gene Name
cadherin 8
Gene Aliases
Nbla04261
UniGene
Hs.368322
Gene Chromosome Location
Chr.16: 61640435-62036835 on Build GRCh38
Transcripts
NM_001796.4 XM_005255760.3 XM_017022829.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
972103 c.1013G>T p.R338M Chr.16: 61820936-61820936 on Build GRCh38
6859922 c.1023+7T>A p.? Chr.16: 61820919-61820919 on Build GRCh38
342849 c.1017A>T p.L339L Chr.16: 61820932-61820932 on Build GRCh38
1563571 c.993G>A p.Q331Q Chr.16: 61820956-61820956 on Build GRCh38
6294601 c.1005C>A p.G335G Chr.16: 61820944-61820944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554898298 Chr.16: 61820902-61820902 on Build GRCh38
rs762478021 Chr.16: 61820961-61820961 on Build GRCh38
rs755278254 Chr.16: 61820895-61820895 on Build GRCh38
rs1051010674 Chr.16: 61820803-61820803 on Build GRCh38
rs569694319 Chr.16: 61820791-61820791 on Build GRCh38
rs574584035 Chr.16: 61820955-61820955 on Build GRCh38
rs745340656 Chr.16: 61820946-61820946 on Build GRCh38
rs779959899 Chr.16: 61820869-61820869 on Build GRCh38
rs776552459 Chr.16: 61820937-61820937 on Build GRCh38
rs750470751 Chr.16: 61820881-61820881 on Build GRCh38
rs202226552 Chr.16: 61820934-61820934 on Build GRCh38
rs147543181 Chr.16: 61820806-61820806 on Build GRCh38
rs922007037 Chr.16: 61820853-61820853 on Build GRCh38
rs769505518 Chr.16: 61820841-61820841 on Build GRCh38
rs541365056 Chr.16: 61820867-61820867 on Build GRCh38
rs773437364 Chr.16: 61820848-61820848 on Build GRCh38
rs34935423 Chr.16: 61820925-61820926 on Build GRCh38
rs947375571 Chr.16: 61820839-61820839 on Build GRCh38
rs756269005 Chr.16: 61820888-61820888 on Build GRCh38
rs558552187 Chr.16: 61820847-61820847 on Build GRCh38
rs116551854 Chr.16: 61820855-61820855 on Build GRCh38
rs376909472 Chr.16: 61820921-61820921 on Build GRCh38
rs749498653 Chr.16: 61820891-61820891 on Build GRCh38
rs932180293 Chr.16: 61820772-61820772 on Build GRCh38
rs775029800 Chr.16: 61820956-61820956 on Build GRCh38
rs770762784 Chr.16: 61820915-61820915 on Build GRCh38
rs777400234 Chr.16: 61820897-61820897 on Build GRCh38
rs780385659 Chr.16: 61820890-61820890 on Build GRCh38
rs190032811 Chr.16: 61820785-61820785 on Build GRCh38
rs768192713 Chr.16: 61820876-61820876 on Build GRCh38

View Full Product Details