Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40252244-40252495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUB1B-PAK6
Entrez Gene ID
106821730
Gene Name
BUB1B-PAK6 readthrough
Gene Aliases
-
UniGene
Hs.732375
Gene Chromosome Location
Chr.15: 40217428-40277487 on Build GRCh38
Transcripts
NM_001128628.2 NM_001128629.2 NM_001039905.2 NM_001276718.1 NM_020168.5

Gene Symbol
C15orf56
Entrez Gene ID
644809
Gene Name
chromosome 15 open reading frame 56
Gene Aliases
-
UniGene
Hs.631718
Gene Chromosome Location
Chr.15: 40250664-40252909 on Build GRCh38
Transcripts
NM_001128628.2 NM_001128629.2 NM_001039905.2 NM_001276718.1 NM_020168.5

Gene Symbol
PAK6
Entrez Gene ID
56924
Gene Name
p21 (RAC1) activated kinase 6
Gene Aliases
PAK5
UniGene
Hs.732375
Gene Chromosome Location
Chr.15: 40239091-40277487 on Build GRCh38
Transcripts
NM_001128628.2 NM_001128629.2 NM_001039905.2 NM_001276718.1 NM_020168.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5564398 c.394C>T p.R132W Chr.15: 40252377-40252377 on Build GRCh38
4884006 c.440C>A p.T147K Chr.15: 40252331-40252331 on Build GRCh38
4853835 c.457C>A p.R153S Chr.15: 40252314-40252314 on Build GRCh38
6425611 c.430T>C p.C144R Chr.15: 40252341-40252341 on Build GRCh38
3815955 c.347C>G p.S116C Chr.15: 40252424-40252424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562091767 Chr.15: 40252336-40252336 on Build GRCh38
rs370638959 Chr.15: 40252306-40252306 on Build GRCh38
rs764361427 Chr.15: 40252332-40252333 on Build GRCh38
rs55863440 Chr.15: 40252416-40252416 on Build GRCh38
rs766770622 Chr.15: 40252458-40252458 on Build GRCh38
rs752341297 Chr.15: 40252462-40252462 on Build GRCh38
rs764313551 Chr.15: 40252323-40252323 on Build GRCh38
rs772010836 Chr.15: 40252404-40252404 on Build GRCh38
rs747589956 Chr.15: 40252282-40252282 on Build GRCh38
rs55799438 Chr.15: 40252292-40252292 on Build GRCh38
rs117614422 Chr.15: 40252330-40252330 on Build GRCh38
rs893750258 Chr.15: 40252309-40252309 on Build GRCh38
rs777674535 Chr.15: 40252284-40252284 on Build GRCh38
rs906365628 Chr.15: 40252357-40252357 on Build GRCh38
rs763281874 Chr.15: 40252444-40252444 on Build GRCh38
rs775863490 Chr.15: 40252311-40252311 on Build GRCh38
rs758795556 Chr.15: 40252348-40252348 on Build GRCh38
rs749252121 Chr.15: 40252287-40252287 on Build GRCh38
rs550607348 Chr.15: 40252396-40252396 on Build GRCh38
rs780775279 Chr.15: 40252352-40252352 on Build GRCh38
rs770713785 Chr.15: 40252389-40252389 on Build GRCh38
rs750689116 Chr.15: 40252341-40252341 on Build GRCh38
rs1053373850 Chr.15: 40252301-40252301 on Build GRCh38
rs868858964 Chr.15: 40252313-40252313 on Build GRCh38
rs761960069 Chr.15: 40252426-40252426 on Build GRCh38
rs566276564 Chr.15: 40252386-40252386 on Build GRCh38
rs944723251 Chr.15: 40252346-40252346 on Build GRCh38
rs896811391 Chr.15: 40252459-40252459 on Build GRCh38
rs55974545 Chr.15: 40252319-40252319 on Build GRCh38
rs376088458 Chr.15: 40252391-40252391 on Build GRCh38
rs754536400 Chr.15: 40252261-40252261 on Build GRCh38
rs527893521 Chr.15: 40252338-40252338 on Build GRCh38
rs772386466 Chr.15: 40252310-40252310 on Build GRCh38
rs759399859 Chr.15: 40252308-40252308 on Build GRCh38
rs765624068 Chr.15: 40252437-40252437 on Build GRCh38
rs775828779 Chr.15: 40252412-40252412 on Build GRCh38
rs780852906 Chr.15: 40252268-40252268 on Build GRCh38
rs769306575 Chr.15: 40252283-40252283 on Build GRCh38
rs1036200308 Chr.15: 40252372-40252372 on Build GRCh38
rs750962796 Chr.15: 40252438-40252438 on Build GRCh38
rs761049600 Chr.15: 40252414-40252414 on Build GRCh38
rs776518212 Chr.15: 40252425-40252425 on Build GRCh38
rs752396383 Chr.15: 40252354-40252354 on Build GRCh38
rs200940821 Chr.15: 40252367-40252367 on Build GRCh38
rs983430667 Chr.15: 40252470-40252470 on Build GRCh38
rs749847018 Chr.15: 40252363-40252363 on Build GRCh38
rs1015338196 Chr.15: 40252448-40252448 on Build GRCh38
rs546368665 Chr.15: 40252376-40252376 on Build GRCh38
rs969896778 Chr.15: 40252291-40252291 on Build GRCh38

View Full Product Details