Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 57869011-57869284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUEDC1
Entrez Gene ID
404093
Gene Name
CUE domain containing 1
Gene Aliases
-
UniGene
Hs.46679
Gene Chromosome Location
Chr.17: 57861243-57955323 on Build GRCh38
Transcripts
NM_001271875.1 NM_001292025.1 XM_006721900.2 XM_011524812.2 XM_017024640.1 XM_017024641.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5055626 c.880G>A p.A294T Chr.17: 57869182-57869182 on Build GRCh38
3970181 c.896C>T p.S299F Chr.17: 57869166-57869166 on Build GRCh38
981698 c.929A>G p.H310R Chr.17: 57869133-57869133 on Build GRCh38
4426248 c.879C>T p.D293D Chr.17: 57869183-57869183 on Build GRCh38
7363320 c.916G>T p.D306Y Chr.17: 57869146-57869146 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766335129 Chr.17: 57869174-57869174 on Build GRCh38
rs568298332 Chr.17: 57869238-57869238 on Build GRCh38
rs560013193 Chr.17: 57869101-57869101 on Build GRCh38
rs779540205 Chr.17: 57869197-57869197 on Build GRCh38
rs773411131 Chr.17: 57869077-57869077 on Build GRCh38
rs79637497 Chr.17: 57869201-57869201 on Build GRCh38
rs368994819 Chr.17: 57869188-57869188 on Build GRCh38
rs373522720 Chr.17: 57869227-57869227 on Build GRCh38
rs373753016 Chr.17: 57869213-57869213 on Build GRCh38
rs759539763 Chr.17: 57869223-57869223 on Build GRCh38
rs747616651 Chr.17: 57869081-57869082 on Build GRCh38
rs1048446094 Chr.17: 57869182-57869182 on Build GRCh38
rs369548166 Chr.17: 57869207-57869207 on Build GRCh38
rs560291011 Chr.17: 57869165-57869165 on Build GRCh38
rs371099612 Chr.17: 57869186-57869186 on Build GRCh38
rs149814069 Chr.17: 57869181-57869181 on Build GRCh38
rs927867335 Chr.17: 57869111-57869111 on Build GRCh38
rs761592191 Chr.17: 57869135-57869135 on Build GRCh38
rs762829876 Chr.17: 57869166-57869166 on Build GRCh38
rs527334239 Chr.17: 57869178-57869178 on Build GRCh38
rs752557322 Chr.17: 57869226-57869226 on Build GRCh38
rs769511161 Chr.17: 57869212-57869212 on Build GRCh38
rs749964639 Chr.17: 57869157-57869157 on Build GRCh38
rs375512742 Chr.17: 57869205-57869205 on Build GRCh38
rs749751641 Chr.17: 57869090-57869090 on Build GRCh38
rs531745800 Chr.17: 57869035-57869035 on Build GRCh38
rs758633825 Chr.17: 57869236-57869236 on Build GRCh38
rs375413839 Chr.17: 57869219-57869219 on Build GRCh38
rs779199890 Chr.17: 57869240-57869240 on Build GRCh38
rs371706527 Chr.17: 57869072-57869072 on Build GRCh38
rs529119429 Chr.17: 57869254-57869254 on Build GRCh38
rs898007308 Chr.17: 57869040-57869040 on Build GRCh38
rs769314844 Chr.17: 57869102-57869102 on Build GRCh38
rs8066051 Chr.17: 57869074-57869074 on Build GRCh38
rs116654850 Chr.17: 57869183-57869183 on Build GRCh38
rs751542943 Chr.17: 57869177-57869177 on Build GRCh38
rs778020217 Chr.17: 57869235-57869235 on Build GRCh38
rs751489734 Chr.17: 57869222-57869222 on Build GRCh38
rs749406977 Chr.17: 57869194-57869194 on Build GRCh38
rs201822381 Chr.17: 57869112-57869112 on Build GRCh38
rs772649310 Chr.17: 57869245-57869245 on Build GRCh38
rs977818323 Chr.17: 57869102-57869102 on Build GRCh38
rs776133007 Chr.17: 57869128-57869128 on Build GRCh38
rs376755509 Chr.17: 57869242-57869242 on Build GRCh38
rs547565691 Chr.17: 57869256-57869256 on Build GRCh38
rs764586502 Chr.17: 57869136-57869136 on Build GRCh38
rs8067058 Chr.17: 57869034-57869034 on Build GRCh38
rs759947730 Chr.17: 57869115-57869115 on Build GRCh38
rs1037076078 Chr.17: 57869203-57869203 on Build GRCh38
rs752861308 Chr.17: 57869187-57869187 on Build GRCh38
rs376505650 Chr.17: 57869220-57869220 on Build GRCh38
rs767974079 Chr.17: 57869123-57869123 on Build GRCh38
rs777662888 Chr.17: 57869192-57869192 on Build GRCh38

View Full Product Details