Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19909779-19910052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP10
Entrez Gene ID
11216
Gene Name
A-kinase anchoring protein 10
Gene Aliases
AKAP-10, D-AKAP-2, D-AKAP2, PRKA10
UniGene
Hs.642676
Gene Chromosome Location
Chr.17: 19904452-19977856 on Build GRCh38
Transcripts
NM_007202.3 XM_006721432.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5385849 c.1839C>T p.S613S Chr.17: 19909974-19909974 on Build GRCh38
6523363 c.1882G>T p.D628Y Chr.17: 19909931-19909931 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs548983236 Chr.17: 19909856-19909856 on Build GRCh38
rs764949322 Chr.17: 19909877-19909877 on Build GRCh38
rs1026097765 Chr.17: 19909964-19909964 on Build GRCh38
rs768794508 Chr.17: 19910000-19910000 on Build GRCh38
rs763908108 Chr.17: 19909914-19909914 on Build GRCh38
rs769377338 Chr.17: 19910019-19910019 on Build GRCh38
rs535264302 Chr.17: 19909864-19909864 on Build GRCh38
rs756229283 Chr.17: 19909983-19909983 on Build GRCh38
rs143579831 Chr.17: 19909974-19909974 on Build GRCh38
rs1016070591 Chr.17: 19909858-19909858 on Build GRCh38
rs779274245 Chr.17: 19910006-19910006 on Build GRCh38
rs748921419 Chr.17: 19909896-19909896 on Build GRCh38
rs367737225 Chr.17: 19909873-19909873 on Build GRCh38
rs780624161 Chr.17: 19909981-19909981 on Build GRCh38
rs751316979 Chr.17: 19909946-19909946 on Build GRCh38
rs748541275 Chr.17: 19910007-19910007 on Build GRCh38
rs772698072 Chr.17: 19910012-19910012 on Build GRCh38
rs750498515 Chr.17: 19909976-19909976 on Build GRCh38
rs757128209 Chr.17: 19909962-19909962 on Build GRCh38
rs749481530 Chr.17: 19909999-19909999 on Build GRCh38
rs773890874 Chr.17: 19910018-19910018 on Build GRCh38
rs374798432 Chr.17: 19909882-19909882 on Build GRCh38
rs377275401 Chr.17: 19909910-19909910 on Build GRCh38
rs920005292 Chr.17: 19909845-19909845 on Build GRCh38
rs905802290 Chr.17: 19909863-19909863 on Build GRCh38
rs931165606 Chr.17: 19909855-19909855 on Build GRCh38
rs113754472 Chr.17: 19909889-19909889 on Build GRCh38
rs780119071 Chr.17: 19909987-19909987 on Build GRCh38
rs987484422 Chr.17: 19910003-19910003 on Build GRCh38
rs143635673 Chr.17: 19909905-19909905 on Build GRCh38

View Full Product Details