Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44197522-44197785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSER1
Entrez Gene ID
51526
Gene Name
oxidative stress responsive serine rich 1
Gene Aliases
C20orf111, HSPC207, Osr1, Perit1, dJ1183I21.1
UniGene
Hs.75798
Gene Chromosome Location
Chr.20: 44195939-44217229 on Build GRCh38
Transcripts
NM_016470.7 XM_011528851.1 XM_011528853.2 XM_011528854.2 XM_017027873.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7109979 c.370T>C p.S124P Chr.20: 44197561-44197561 on Build GRCh38
1130629 c.292A>G p.I98V Chr.20: 44197639-44197639 on Build GRCh38
7138129 c.296C>T p.A99V Chr.20: 44197635-44197635 on Build GRCh38
4793595 c.231G>A p.Q77Q Chr.20: 44197700-44197700 on Build GRCh38
126584 c.238C>T p.R80* Chr.20: 44197693-44197693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529359405 Chr.20: 44197610-44197610 on Build GRCh38
rs765374173 Chr.20: 44197649-44197649 on Build GRCh38
rs903491002 Chr.20: 44197704-44197704 on Build GRCh38
rs761920502 Chr.20: 44197562-44197562 on Build GRCh38
rs143943850 Chr.20: 44197635-44197635 on Build GRCh38
rs765441301 Chr.20: 44197569-44197569 on Build GRCh38
rs896277871 Chr.20: 44197759-44197759 on Build GRCh38
rs1051236097 Chr.20: 44197631-44197631 on Build GRCh38
rs756944504 Chr.20: 44197722-44197722 on Build GRCh38
rs763498683 Chr.20: 44197695-44197695 on Build GRCh38
rs374753400 Chr.20: 44197597-44197597 on Build GRCh38
rs781639706 Chr.20: 44197749-44197749 on Build GRCh38
rs755382758 Chr.20: 44197585-44197585 on Build GRCh38
rs756709729 Chr.20: 44197598-44197598 on Build GRCh38
rs768525162 Chr.20: 44197751-44197751 on Build GRCh38
rs1044386435 Chr.20: 44197689-44197689 on Build GRCh38
rs562498505 Chr.20: 44197719-44197719 on Build GRCh38
rs9346 Chr.20: 44197710-44197710 on Build GRCh38
rs752110025 Chr.20: 44197698-44197698 on Build GRCh38
rs756835305 Chr.20: 44197743-44197748 on Build GRCh38
rs142485619 Chr.20: 44197607-44197607 on Build GRCh38
rs372148209 Chr.20: 44197557-44197557 on Build GRCh38
rs753468232 Chr.20: 44197720-44197720 on Build GRCh38
rs201888626 Chr.20: 44197574-44197574 on Build GRCh38
rs972827935 Chr.20: 44197583-44197583 on Build GRCh38
rs775345649 Chr.20: 44197616-44197616 on Build GRCh38
rs540214632 Chr.20: 44197554-44197554 on Build GRCh38
rs942924692 Chr.20: 44197642-44197642 on Build GRCh38
rs199512438 Chr.20: 44197645-44197645 on Build GRCh38
rs758864876 Chr.20: 44197573-44197573 on Build GRCh38
rs776605311 Chr.20: 44197758-44197758 on Build GRCh38
rs747001419 Chr.20: 44197749-44197749 on Build GRCh38
rs368625214 Chr.20: 44197760-44197760 on Build GRCh38
rs748114723 Chr.20: 44197634-44197634 on Build GRCh38
rs745428737 Chr.20: 44197735-44197735 on Build GRCh38
rs750917568 Chr.20: 44197570-44197570 on Build GRCh38
rs907867807 Chr.20: 44197636-44197636 on Build GRCh38
rs373020907 Chr.20: 44197696-44197696 on Build GRCh38
rs11274 Chr.20: 44197739-44197739 on Build GRCh38
rs775189988 Chr.20: 44197552-44197552 on Build GRCh38
rs768536425 Chr.20: 44197559-44197559 on Build GRCh38
rs774678227 Chr.20: 44197580-44197580 on Build GRCh38
rs773228353 Chr.20: 44197672-44197672 on Build GRCh38
rs147662241 Chr.20: 44197728-44197728 on Build GRCh38

View Full Product Details