Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29816770-29817015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAGR1
Entrez Gene ID
79447
Gene Name
PAXIP1 associated glutamate rich protein 1
Gene Aliases
C16orf53, GAS, PA1
UniGene
Hs.702841
Gene Chromosome Location
Chr.16: 29816207-29822495 on Build GRCh38
Transcripts
NM_024516.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5955483 c.303T>A p.D101E Chr.16: 29816828-29816828 on Build GRCh38
5951864 c.409C>T p.P137S Chr.16: 29816934-29816934 on Build GRCh38
6496646 c.407C>T p.P136L Chr.16: 29816932-29816932 on Build GRCh38
6411836 c.451G>T p.E151* Chr.16: 29816976-29816976 on Build GRCh38
5669035 c.357G>A p.L119L Chr.16: 29816882-29816882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867495905 Chr.16: 29816955-29816955 on Build GRCh38
rs758338974 Chr.16: 29816956-29816956 on Build GRCh38
rs1036699712 Chr.16: 29816851-29816851 on Build GRCh38
rs778813478 Chr.16: 29816817-29816817 on Build GRCh38
rs780031691 Chr.16: 29816962-29816962 on Build GRCh38
rs534165002 Chr.16: 29816848-29816848 on Build GRCh38
rs769365806 Chr.16: 29816975-29816975 on Build GRCh38
rs377615776 Chr.16: 29816901-29816901 on Build GRCh38
rs751503327 Chr.16: 29816965-29816965 on Build GRCh38
rs753983836 Chr.16: 29816911-29816911 on Build GRCh38
rs867151087 Chr.16: 29816926-29816926 on Build GRCh38
rs960719579 Chr.16: 29816987-29816987 on Build GRCh38
rs745681136 Chr.16: 29816825-29816825 on Build GRCh38
rs779860747 Chr.16: 29816843-29816843 on Build GRCh38
rs369349605 Chr.16: 29816865-29816865 on Build GRCh38
rs759895360 Chr.16: 29816887-29816887 on Build GRCh38
rs374459711 Chr.16: 29816941-29816941 on Build GRCh38
rs768003848 Chr.16: 29816896-29816896 on Build GRCh38
rs757085801 Chr.16: 29816811-29816811 on Build GRCh38
rs758354996 Chr.16: 29816827-29816827 on Build GRCh38
rs909600238 Chr.16: 29816815-29816815 on Build GRCh38
rs747798431 Chr.16: 29816872-29816872 on Build GRCh38
rs769455895 Chr.16: 29816879-29816879 on Build GRCh38
rs752751554 Chr.16: 29816790-29816790 on Build GRCh38
rs754054325 Chr.16: 29816802-29816802 on Build GRCh38
rs1030986454 Chr.16: 29816980-29816980 on Build GRCh38
rs918289522 Chr.16: 29816862-29816862 on Build GRCh38
rs572999151 Chr.16: 29816844-29816844 on Build GRCh38
rs373200391 Chr.16: 29816884-29816884 on Build GRCh38
rs370072946 Chr.16: 29816912-29816912 on Build GRCh38
rs750176614 Chr.16: 29816954-29816954 on Build GRCh38
rs558886856 Chr.16: 29816903-29816903 on Build GRCh38
rs375215655 Chr.16: 29816799-29816799 on Build GRCh38
rs764118426 Chr.16: 29816910-29816910 on Build GRCh38
rs754482731 Chr.16: 29816968-29816968 on Build GRCh38
rs780516799 Chr.16: 29816970-29816970 on Build GRCh38
rs777464299 Chr.16: 29816989-29816989 on Build GRCh38
rs868558474 Chr.16: 29816899-29816899 on Build GRCh38
rs1050989106 Chr.16: 29816880-29816880 on Build GRCh38

View Full Product Details