Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 59475875-59476116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
L3HYPDH
Entrez Gene ID
112849
Gene Name
trans-L-3-hydroxyproline dehydratase
Gene Aliases
C14orf149
UniGene
Hs.729061
Gene Chromosome Location
Chr.14: 59465545-59484438 on Build GRCh38
Transcripts
NM_144581.1 XM_005267298.3 XM_011536386.2 XM_011536387.1 XM_011536388.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6076025 c.808A>T p.R270* Chr.14: 59476000-59476000 on Build GRCh38
3401372 c.791C>T p.A264V Chr.14: 59476102-59476102 on Build GRCh38
1128237 c.798A>C p.E266D Chr.14: 59476095-59476095 on Build GRCh38
6567914 c.921C>T p.F307F Chr.14: 59475887-59475887 on Build GRCh38
2246462 c.802G>A p.V268I Chr.14: 59476006-59476006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746854388 Chr.14: 59476054-59476054 on Build GRCh38
rs746012365 Chr.14: 59475913-59475913 on Build GRCh38
rs150191667 Chr.14: 59475956-59475956 on Build GRCh38
rs375970299 Chr.14: 59475970-59475970 on Build GRCh38
rs111662029 Chr.14: 59476027-59476027 on Build GRCh38
rs754750495 Chr.14: 59476056-59476056 on Build GRCh38
rs757399959 Chr.14: 59475901-59475901 on Build GRCh38
rs963397311 Chr.14: 59476044-59476044 on Build GRCh38
rs781168532 Chr.14: 59476070-59476070 on Build GRCh38
rs761367011 Chr.14: 59475928-59475928 on Build GRCh38
rs771916154 Chr.14: 59475920-59475920 on Build GRCh38
rs746978380 Chr.14: 59475924-59475924 on Build GRCh38
rs373034520 Chr.14: 59476053-59476053 on Build GRCh38
rs867528098 Chr.14: 59476007-59476007 on Build GRCh38
rs369002443 Chr.14: 59476024-59476024 on Build GRCh38
rs758507597 Chr.14: 59476043-59476043 on Build GRCh38
rs769178897 Chr.14: 59475946-59475946 on Build GRCh38
rs929839369 Chr.14: 59476086-59476086 on Build GRCh38
rs748009012 Chr.14: 59476071-59476071 on Build GRCh38
rs778802447 Chr.14: 59476016-59476016 on Build GRCh38
rs761014280 Chr.14: 59476059-59476059 on Build GRCh38
rs779261976 Chr.14: 59475906-59475906 on Build GRCh38
rs757364366 Chr.14: 59476000-59476000 on Build GRCh38
rs879579189 Chr.14: 59475940-59475940 on Build GRCh38
rs1025061389 Chr.14: 59475919-59475919 on Build GRCh38
rs371673690 Chr.14: 59475969-59475969 on Build GRCh38
rs768619597 Chr.14: 59475925-59475925 on Build GRCh38
rs762453312 Chr.14: 59475958-59475958 on Build GRCh38
rs754393738 Chr.14: 59475987-59475987 on Build GRCh38
rs779662529 Chr.14: 59475921-59475921 on Build GRCh38
rs769357881 Chr.14: 59476074-59476074 on Build GRCh38
rs757047105 Chr.14: 59476029-59476030 on Build GRCh38
rs776527761 Chr.14: 59475927-59475927 on Build GRCh38
rs767958121 Chr.14: 59475964-59475964 on Build GRCh38

View Full Product Details