Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 73704109-73704380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD276
Entrez Gene ID
80381
Gene Name
CD276 molecule
Gene Aliases
4Ig-B7-H3, B7-H3, B7H3, B7RP-2
UniGene
Hs.744915
Gene Chromosome Location
Chr.15: 73683966-73714518 on Build GRCh38
Transcripts
NM_001024736.1 NM_025240.2 XM_005254700.4 XM_011522095.2 XM_011522096.2 XM_017022638.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
701271 c.1160G>A p.R387Q Chr.15: 73704263-73704263 on Build GRCh38
1374456 c.1141A>G p.I381V Chr.15: 73704244-73704244 on Build GRCh38
701270 c.1230G>T p.T410T Chr.15: 73704333-73704333 on Build GRCh38
4056749 c.1233G>A p.S411S Chr.15: 73704336-73704336 on Build GRCh38
4056748 c.1121G>A p.R374Q Chr.15: 73704224-73704224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375906969 Chr.15: 73704129-73704129 on Build GRCh38
rs779619317 Chr.15: 73704150-73704151 on Build GRCh38
rs982390898 Chr.15: 73704262-73704262 on Build GRCh38
rs776236542 Chr.15: 73704336-73704336 on Build GRCh38
rs748073003 Chr.15: 73704328-73704330 on Build GRCh38
rs941216324 Chr.15: 73704127-73704127 on Build GRCh38
rs763468689 Chr.15: 73704189-73704189 on Build GRCh38
rs759855936 Chr.15: 73704183-73704183 on Build GRCh38
rs772221210 Chr.15: 73704155-73704155 on Build GRCh38
rs758384094 Chr.15: 73704268-73704268 on Build GRCh38
rs747184452 Chr.15: 73704163-73704163 on Build GRCh38
rs919519613 Chr.15: 73704358-73704361 on Build GRCh38
rs377370844 Chr.15: 73704171-73704171 on Build GRCh38
rs374890808 Chr.15: 73704257-73704257 on Build GRCh38
rs150186392 Chr.15: 73704263-73704263 on Build GRCh38
rs1020175441 Chr.15: 73704279-73704279 on Build GRCh38
rs762905746 Chr.15: 73704349-73704349 on Build GRCh38
rs766950718 Chr.15: 73704194-73704194 on Build GRCh38
rs775922140 Chr.15: 73704186-73704186 on Build GRCh38
rs368813244 Chr.15: 73704237-73704237 on Build GRCh38
rs141446768 Chr.15: 73704258-73704258 on Build GRCh38
rs777947795 Chr.15: 73704232-73704232 on Build GRCh38
rs145827704 Chr.15: 73704236-73704236 on Build GRCh38
rs759060595 Chr.15: 73704339-73704339 on Build GRCh38
rs150877011 Chr.15: 73704360-73704360 on Build GRCh38
rs376481613 Chr.15: 73704348-73704348 on Build GRCh38
rs781015787 Chr.15: 73704321-73704323 on Build GRCh38
rs372124791 Chr.15: 73704335-73704335 on Build GRCh38
rs926489834 Chr.15: 73704319-73704319 on Build GRCh38
rs369412356 Chr.15: 73704324-73704324 on Build GRCh38
rs976534600 Chr.15: 73704274-73704274 on Build GRCh38
rs774907009 Chr.15: 73704332-73704332 on Build GRCh38
rs764107033 Chr.15: 73704359-73704359 on Build GRCh38
rs138724355 Chr.15: 73704316-73704316 on Build GRCh38
rs185182962 Chr.15: 73704248-73704248 on Build GRCh38
rs779785470 Chr.15: 73704142-73704142 on Build GRCh38
rs962915686 Chr.15: 73704266-73704266 on Build GRCh38
rs765772947 Chr.15: 73704185-73704185 on Build GRCh38
rs761420087 Chr.15: 73704156-73704156 on Build GRCh38
rs778781915 Chr.15: 73704148-73704148 on Build GRCh38
rs771297187 Chr.15: 73704169-73704169 on Build GRCh38
rs373656928 Chr.15: 73704147-73704147 on Build GRCh38
rs748030700 Chr.15: 73704225-73704225 on Build GRCh38
rs149360961 Chr.15: 73704325-73704325 on Build GRCh38
rs753639536 Chr.15: 73704209-73704209 on Build GRCh38
rs776781918 Chr.15: 73704243-73704243 on Build GRCh38
rs751682061 Chr.15: 73704288-73704288 on Build GRCh38
rs201073171 Chr.15: 73704312-73704312 on Build GRCh38
rs772688844 Chr.15: 73704254-73704254 on Build GRCh38
rs757341580 Chr.15: 73704294-73704294 on Build GRCh38
rs752558512 Chr.15: 73704264-73704264 on Build GRCh38
rs200610531 Chr.15: 73704259-73704259 on Build GRCh38
rs754785892 Chr.15: 73704223-73704223 on Build GRCh38
rs375111114 Chr.15: 73704226-73704226 on Build GRCh38
rs749062091 Chr.15: 73704143-73704143 on Build GRCh38
rs370885634 Chr.15: 73704204-73704204 on Build GRCh38
rs577104872 Chr.15: 73704160-73704160 on Build GRCh38
rs143706609 Chr.15: 73704333-73704333 on Build GRCh38
rs371919678 Chr.15: 73704138-73704138 on Build GRCh38
rs143255170 Chr.15: 73704249-73704249 on Build GRCh38
rs375349822 Chr.15: 73704295-73704295 on Build GRCh38
rs778780413 Chr.15: 73704224-73704224 on Build GRCh38
rs201851630 Chr.15: 73704165-73704165 on Build GRCh38
rs141658038 Chr.15: 73704220-73704220 on Build GRCh38
rs754783784 Chr.15: 73704290-73704290 on Build GRCh38
rs140663039 Chr.15: 73704196-73704196 on Build GRCh38
rs769999909 Chr.15: 73704244-73704244 on Build GRCh38
rs758439759 Chr.15: 73704228-73704228 on Build GRCh38
rs777894870 Chr.15: 73704280-73704280 on Build GRCh38
rs775931869 Chr.15: 73704245-73704245 on Build GRCh38
rs751224675 Chr.15: 73704163-73704164 on Build GRCh38
rs748243761 Chr.15: 73704152-73704152 on Build GRCh38

View Full Product Details