Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 13417077-13417330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD10
Entrez Gene ID
84280
Gene Name
BTB domain containing 10
Gene Aliases
GMRP-1, GMRP1
UniGene
Hs.332382
Gene Chromosome Location
Chr.11: 13387996-13463297 on Build GRCh38
Transcripts
NM_001297741.1 NM_001297742.1 NM_032320.6 XM_017018405.1 XM_017018406.1 XM_017018407.1 XM_017018408.1 XM_017018409.1 XM_017018410.1 XM_017018411.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2090513 c.676C>T p.R226* Chr.11: 13417169-13417169 on Build GRCh38
6890980 c.620G>A p.R207Q Chr.11: 13417225-13417225 on Build GRCh38
7090190 c.654G>T p.E218D Chr.11: 13417191-13417191 on Build GRCh38
7427290 c.669T>C p.T223T Chr.11: 13417176-13417176 on Build GRCh38
6405159 c.658A>C p.I220L Chr.11: 13417187-13417187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770882478 Chr.11: 13417219-13417219 on Build GRCh38
rs760365769 Chr.11: 13417120-13417120 on Build GRCh38
rs201616431 Chr.11: 13417164-13417164 on Build GRCh38
rs377317797 Chr.11: 13417168-13417168 on Build GRCh38
rs777903984 Chr.11: 13417139-13417139 on Build GRCh38
rs774014312 Chr.11: 13417266-13417266 on Build GRCh38
rs765553013 Chr.11: 13417296-13417296 on Build GRCh38
rs764638325 Chr.11: 13417144-13417144 on Build GRCh38
rs760111658 Chr.11: 13417283-13417283 on Build GRCh38
rs751848050 Chr.11: 13417150-13417151 on Build GRCh38
rs745798831 Chr.11: 13417253-13417253 on Build GRCh38
rs762174876 Chr.11: 13417143-13417143 on Build GRCh38
rs139827531 Chr.11: 13417249-13417249 on Build GRCh38
rs915063868 Chr.11: 13417208-13417208 on Build GRCh38
rs536418438 Chr.11: 13417244-13417244 on Build GRCh38
rs150841566 Chr.11: 13417166-13417166 on Build GRCh38
rs377624520 Chr.11: 13417288-13417288 on Build GRCh38
rs768235133 Chr.11: 13417285-13417285 on Build GRCh38
rs765681805 Chr.11: 13417150-13417150 on Build GRCh38
rs749042411 Chr.11: 13417178-13417178 on Build GRCh38
rs761940876 Chr.11: 13417261-13417261 on Build GRCh38
rs112373983 Chr.11: 13417174-13417174 on Build GRCh38
rs771964612 Chr.11: 13417260-13417260 on Build GRCh38
rs369889215 Chr.11: 13417145-13417145 on Build GRCh38
rs763805720 Chr.11: 13417122-13417122 on Build GRCh38
rs774915987 Chr.11: 13417119-13417119 on Build GRCh38
rs946573049 Chr.11: 13417210-13417210 on Build GRCh38
rs750704814 Chr.11: 13417292-13417292 on Build GRCh38
rs960126769 Chr.11: 13417238-13417238 on Build GRCh38
rs867833527 Chr.11: 13417263-13417263 on Build GRCh38
rs750968552 Chr.11: 13417160-13417160 on Build GRCh38
rs372858468 Chr.11: 13417206-13417206 on Build GRCh38
rs375488801 Chr.11: 13417126-13417126 on Build GRCh38
rs865851316 Chr.11: 13417185-13417185 on Build GRCh38
rs554936945 Chr.11: 13417287-13417287 on Build GRCh38

View Full Product Details