Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207699122-207699375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CR1L
Entrez Gene ID
1379
Gene Name
complement component 3b/4b receptor 1-like
Gene Aliases
-
UniGene
Hs.628568
Gene Chromosome Location
Chr.1: 207645113-207723691 on Build GRCh38
Transcripts
NM_175710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6460003 c.1182G>C p.L394F Chr.1: 207699228-207699228 on Build GRCh38
3746757 c.1228A>G p.N410D Chr.1: 207699250-207699250 on Build GRCh38
5522432 c.1202G>A p.W401* Chr.1: 207699248-207699248 on Build GRCh38
3864343 c.1157G>A p.G386D Chr.1: 207699203-207699203 on Build GRCh38
6460004 c.1206G>C p.L402F Chr.1: 207699228-207699228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545020633 Chr.1: 207699218-207699218 on Build GRCh38
rs762330333 Chr.1: 207699270-207699270 on Build GRCh38
rs773366212 Chr.1: 207699266-207699266 on Build GRCh38
rs757955316 Chr.1: 207699195-207699195 on Build GRCh38
rs781205737 Chr.1: 207699238-207699238 on Build GRCh38
rs574870537 Chr.1: 207699184-207699184 on Build GRCh38
rs766877113 Chr.1: 207699271-207699271 on Build GRCh38
rs755269019 Chr.1: 207699167-207699167 on Build GRCh38
rs752963448 Chr.1: 207699292-207699292 on Build GRCh38
rs765672897 Chr.1: 207699289-207699289 on Build GRCh38
rs12729569 Chr.1: 207699250-207699250 on Build GRCh38
rs192423219 Chr.1: 207699310-207699310 on Build GRCh38
rs757271882 Chr.1: 207699232-207699232 on Build GRCh38
rs779141978 Chr.1: 207699182-207699182 on Build GRCh38
rs777515448 Chr.1: 207699335-207699335 on Build GRCh38
rs570195079 Chr.1: 207699310-207699310 on Build GRCh38
rs779758176 Chr.1: 207699228-207699228 on Build GRCh38
rs190448008 Chr.1: 207699299-207699299 on Build GRCh38
rs368204114 Chr.1: 207699319-207699319 on Build GRCh38
rs374206906 Chr.1: 207699275-207699275 on Build GRCh38
rs41313728 Chr.1: 207699162-207699162 on Build GRCh38
rs201260912 Chr.1: 207699322-207699322 on Build GRCh38
rs775128887 Chr.1: 207699255-207699255 on Build GRCh38
rs754555671 Chr.1: 207699274-207699274 on Build GRCh38
rs751090378 Chr.1: 207699259-207699259 on Build GRCh38
rs146636186 Chr.1: 207699338-207699338 on Build GRCh38
rs12748243 Chr.1: 207699312-207699312 on Build GRCh38
rs748136682 Chr.1: 207699309-207699309 on Build GRCh38
rs371110227 Chr.1: 207699262-207699262 on Build GRCh38
rs745791420 Chr.1: 207699244-207699244 on Build GRCh38
rs763593797 Chr.1: 207699303-207699303 on Build GRCh38
rs114968622 Chr.1: 207699229-207699229 on Build GRCh38
rs748445817 Chr.1: 207699264-207699264 on Build GRCh38
rs546081271 Chr.1: 207699306-207699306 on Build GRCh38
rs777373419 Chr.1: 207699197-207699197 on Build GRCh38

View Full Product Details