Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 37848541-37848778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRF2
Entrez Gene ID
55290
Gene Name
BRF2, RNA polymerase III transcription initiation factor 50 kDa subunit
Gene Aliases
BRFU, TFIIIB50
UniGene
Hs.709301
Gene Chromosome Location
Chr.8: 37843880-37849913 on Build GRCh38
Transcripts
NM_018310.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6612842 c.173G>A p.S58N Chr.8: 37848637-37848637 on Build GRCh38
6328538 c.181G>A p.E61K Chr.8: 37848629-37848629 on Build GRCh38
1099488 c.210A>G p.Q70Q Chr.8: 37848600-37848600 on Build GRCh38
1099489 c.170G>A p.R57Q Chr.8: 37848640-37848640 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771294099 Chr.8: 37848699-37848699 on Build GRCh38
rs759688798 Chr.8: 37848660-37848660 on Build GRCh38
rs774539967 Chr.8: 37848613-37848613 on Build GRCh38
rs780911415 Chr.8: 37848686-37848686 on Build GRCh38
rs747587959 Chr.8: 37848584-37848584 on Build GRCh38
rs545787925 Chr.8: 37848645-37848645 on Build GRCh38
rs751267936 Chr.8: 37848679-37848679 on Build GRCh38
rs563887611 Chr.8: 37848649-37848649 on Build GRCh38
rs780923859 Chr.8: 37848572-37848572 on Build GRCh38
rs759965518 Chr.8: 37848623-37848623 on Build GRCh38
rs754803351 Chr.8: 37848682-37848682 on Build GRCh38
rs1036705037 Chr.8: 37848576-37848576 on Build GRCh38
rs12541064 Chr.8: 37848749-37848749 on Build GRCh38
rs370316183 Chr.8: 37848689-37848689 on Build GRCh38
rs1003919301 Chr.8: 37848574-37848574 on Build GRCh38
rs968161807 Chr.8: 37848739-37848739 on Build GRCh38
rs765373667 Chr.8: 37848702-37848702 on Build GRCh38
rs190535539 Chr.8: 37848752-37848752 on Build GRCh38
rs768156652 Chr.8: 37848569-37848569 on Build GRCh38
rs376593889 Chr.8: 37848678-37848678 on Build GRCh38
rs897007240 Chr.8: 37848680-37848680 on Build GRCh38
rs753445686 Chr.8: 37848599-37848599 on Build GRCh38
rs748826439 Chr.8: 37848694-37848694 on Build GRCh38
rs754023927 Chr.8: 37848668-37848668 on Build GRCh38
rs767575884 Chr.8: 37848628-37848628 on Build GRCh38
rs747783563 Chr.8: 37848687-37848687 on Build GRCh38
rs760012086 Chr.8: 37848664-37848664 on Build GRCh38
rs200905754 Chr.8: 37848595-37848595 on Build GRCh38
rs1033031215 Chr.8: 37848751-37848751 on Build GRCh38
rs374505515 Chr.8: 37848589-37848589 on Build GRCh38
rs777233389 Chr.8: 37848691-37848691 on Build GRCh38
rs758239469 Chr.8: 37848669-37848669 on Build GRCh38
rs970701297 Chr.8: 37848703-37848703 on Build GRCh38
rs896018342 Chr.8: 37848610-37848610 on Build GRCh38
rs140395188 Chr.8: 37848611-37848611 on Build GRCh38
rs775861500 Chr.8: 37848641-37848641 on Build GRCh38

View Full Product Details