Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 95600892-95601123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSK
Entrez Gene ID
153642
Gene Name
arylsulfatase family member K
Gene Aliases
TSULF
UniGene
Hs.585051
Gene Chromosome Location
Chr.5: 95555108-95605102 on Build GRCh38
Transcripts
NM_198150.2 XM_005271904.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5061273 c.1303A>G p.I435V Chr.5: 95601053-95601053 on Build GRCh38
1487052 c.1159G>C p.E387Q Chr.5: 95600909-95600909 on Build GRCh38
1671761 c.1312C>T p.Q438* Chr.5: 95601062-95601062 on Build GRCh38
3618834 c.1199C>T p.P400L Chr.5: 95600949-95600949 on Build GRCh38
1731453 c.1262C>T p.T421I Chr.5: 95601012-95601012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753742067 Chr.5: 95600990-95600990 on Build GRCh38
rs763396857 Chr.5: 95601039-95601039 on Build GRCh38
rs755905288 Chr.5: 95600971-95600971 on Build GRCh38
rs774511398 Chr.5: 95601089-95601090 on Build GRCh38
rs370552188 Chr.5: 95601009-95601009 on Build GRCh38
rs778009188 Chr.5: 95601004-95601004 on Build GRCh38
rs61733083 Chr.5: 95601040-95601040 on Build GRCh38
rs779878421 Chr.5: 95600976-95600976 on Build GRCh38
rs148460722 Chr.5: 95600962-95600962 on Build GRCh38
rs773863677 Chr.5: 95601041-95601041 on Build GRCh38
rs749379584 Chr.5: 95601001-95601001 on Build GRCh38
rs749670118 Chr.5: 95600964-95600965 on Build GRCh38
rs754905227 Chr.5: 95600993-95600993 on Build GRCh38
rs761369254 Chr.5: 95601044-95601044 on Build GRCh38
rs200763153 Chr.5: 95601071-95601071 on Build GRCh38
rs747031061 Chr.5: 95601008-95601008 on Build GRCh38
rs750127084 Chr.5: 95601064-95601064 on Build GRCh38
rs374765719 Chr.5: 95600925-95600925 on Build GRCh38
rs746068393 Chr.5: 95601028-95601028 on Build GRCh38
rs565377794 Chr.5: 95601080-95601080 on Build GRCh38
rs751029839 Chr.5: 95600920-95600920 on Build GRCh38
rs781630344 Chr.5: 95601014-95601014 on Build GRCh38
rs775870978 Chr.5: 95601034-95601034 on Build GRCh38
rs770102407 Chr.5: 95601029-95601029 on Build GRCh38
rs929760482 Chr.5: 95600943-95600943 on Build GRCh38
rs767008042 Chr.5: 95601056-95601056 on Build GRCh38
rs1054420238 Chr.5: 95600988-95600988 on Build GRCh38
rs892636632 Chr.5: 95600996-95600996 on Build GRCh38
rs868698456 Chr.5: 95600948-95600948 on Build GRCh38
rs1043157588 Chr.5: 95601058-95601058 on Build GRCh38
rs771319401 Chr.5: 95601069-95601070 on Build GRCh38

View Full Product Details