Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6237614-6237883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD9
Entrez Gene ID
928
Gene Name
CD9 molecule
Gene Aliases
BTCC-1, DRAP-27, MIC3, MRP-1, TSPAN-29, TSPAN29
UniGene
Hs.114286
Gene Chromosome Location
Chr.12: 6199844-6238271 on Build GRCh38
Transcripts
NM_001769.3 XM_011521040.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5004631 c.674G>A p.R225H Chr.12: 6237815-6237815 on Build GRCh38
4575653 c.675C>A p.R225R Chr.12: 6237816-6237816 on Build GRCh38
7271652 c.676G>A p.E226K Chr.12: 6237817-6237817 on Build GRCh38
942393 c.675C>T p.R225R Chr.12: 6237816-6237816 on Build GRCh38
942392 c.636C>T p.I212I Chr.12: 6237777-6237777 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017914861 Chr.12: 6237683-6237683 on Build GRCh38
rs751433404 Chr.12: 6237751-6237751 on Build GRCh38
rs17850834 Chr.12: 6237785-6237785 on Build GRCh38
rs748727676 Chr.12: 6237636-6237636 on Build GRCh38
rs11568275 Chr.12: 6237721-6237721 on Build GRCh38
rs11568274 Chr.12: 6237700-6237700 on Build GRCh38
rs985963051 Chr.12: 6237692-6237692 on Build GRCh38
rs771146611 Chr.12: 6237831-6237831 on Build GRCh38
rs775488510 Chr.12: 6237791-6237791 on Build GRCh38
rs950130247 Chr.12: 6237781-6237781 on Build GRCh38
rs1056361459 Chr.12: 6237733-6237733 on Build GRCh38
rs781607093 Chr.12: 6237816-6237816 on Build GRCh38
rs371007109 Chr.12: 6237844-6237844 on Build GRCh38
rs753223924 Chr.12: 6237817-6237817 on Build GRCh38
rs769902776 Chr.12: 6237738-6237738 on Build GRCh38
rs1030073613 Chr.12: 6237654-6237654 on Build GRCh38
rs190218669 Chr.12: 6237815-6237815 on Build GRCh38
rs772068319 Chr.12: 6237789-6237789 on Build GRCh38
rs755552716 Chr.12: 6237714-6237714 on Build GRCh38
rs905280924 Chr.12: 6237839-6237839 on Build GRCh38
rs988979164 Chr.12: 6237773-6237773 on Build GRCh38
rs1038337177 Chr.12: 6237797-6237797 on Build GRCh38
rs116192999 Chr.12: 6237784-6237784 on Build GRCh38
rs753245518 Chr.12: 6237798-6237798 on Build GRCh38
rs756690162 Chr.12: 6237667-6237667 on Build GRCh38
rs146304211 Chr.12: 6237805-6237805 on Build GRCh38
rs913329230 Chr.12: 6237777-6237777 on Build GRCh38
rs777904401 Chr.12: 6237753-6237753 on Build GRCh38
rs748242402 Chr.12: 6237728-6237728 on Build GRCh38
rs771093583 Chr.12: 6237771-6237771 on Build GRCh38
rs976275339 Chr.12: 6237646-6237646 on Build GRCh38
rs149789049 Chr.12: 6237794-6237794 on Build GRCh38
rs957806306 Chr.12: 6237772-6237772 on Build GRCh38
rs749544736 Chr.12: 6237758-6237758 on Build GRCh38
rs569393189 Chr.12: 6237846-6237846 on Build GRCh38
rs974967942 Chr.12: 6237704-6237704 on Build GRCh38
rs779104198 Chr.12: 6237834-6237834 on Build GRCh38
rs925080147 Chr.12: 6237696-6237696 on Build GRCh38
rs751973618 Chr.12: 6237807-6237807 on Build GRCh38
rs760701551 Chr.12: 6237856-6237856 on Build GRCh38
rs760154648 Chr.12: 6237793-6237793 on Build GRCh38
rs953212677 Chr.12: 6237668-6237668 on Build GRCh38
rs181150196 Chr.12: 6237840-6237840 on Build GRCh38
rs201524346 Chr.12: 6237806-6237806 on Build GRCh38
rs777932393 Chr.12: 6237820-6237820 on Build GRCh38
rs913931364 Chr.12: 6237739-6237739 on Build GRCh38
rs117082756 Chr.12: 6237635-6237635 on Build GRCh38

View Full Product Details