Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70449284-70449547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLG3
Entrez Gene ID
1741
Gene Name
discs large MAGUK scaffold protein 3
Gene Aliases
MRX, MRX90, NEDLG, PPP1R82, SAP102, XLMR
UniGene
Hs.721586
Gene Chromosome Location
Chr.X: 70444850-70505490 on Build GRCh38
Transcripts
NM_021120.3 XM_006724625.2 XM_006724626.2 XM_011530883.1

SNP Detail

SNP ID Chromosome Location
rs779238940 Chr.X: 70449477-70449477 on Build GRCh38
rs755292432 Chr.X: 70449468-70449468 on Build GRCh38
rs774169484 Chr.X: 70449395-70449395 on Build GRCh38
rs753026876 Chr.X: 70449486-70449486 on Build GRCh38
rs375246548 Chr.X: 70449523-70449523 on Build GRCh38
rs35852632 Chr.X: 70449442-70449443 on Build GRCh38
rs774974502 Chr.X: 70449374-70449374 on Build GRCh38
rs148860106 Chr.X: 70449415-70449415 on Build GRCh38
rs188475429 Chr.X: 70449355-70449355 on Build GRCh38
rs750311987 Chr.X: 70449410-70449410 on Build GRCh38
rs766625496 Chr.X: 70449413-70449413 on Build GRCh38
rs764262068 Chr.X: 70449496-70449496 on Build GRCh38
rs760528718 Chr.X: 70449412-70449412 on Build GRCh38
rs144225009 Chr.X: 70449379-70449379 on Build GRCh38
rs864321655 Chr.X: 70449482-70449482 on Build GRCh38
rs779586939 Chr.X: 70449518-70449518 on Build GRCh38
rs761291363 Chr.X: 70449460-70449460 on Build GRCh38
rs771412444 Chr.X: 70449512-70449512 on Build GRCh38
rs767373575 Chr.X: 70449409-70449409 on Build GRCh38
rs745382067 Chr.X: 70449322-70449322 on Build GRCh38
rs778146352 Chr.X: 70449493-70449493 on Build GRCh38
rs760909281 Chr.X: 70449489-70449489 on Build GRCh38
rs369293532 Chr.X: 70449528-70449528 on Build GRCh38
rs761625022 Chr.X: 70449397-70449397 on Build GRCh38
rs145494926 Chr.X: 70449385-70449385 on Build GRCh38
rs199982927 Chr.X: 70449481-70449481 on Build GRCh38
rs748763126 Chr.X: 70449519-70449519 on Build GRCh38

View Full Product Details