Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 59984426-59984699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH26
Entrez Gene ID
60437
Gene Name
cadherin 26
Gene Aliases
VR20
UniGene
Hs.729046
Gene Chromosome Location
Chr.20: 59957764-60035629 on Build GRCh38
Transcripts
NM_177980.2 XM_011528969.2 XM_017027994.1 XM_017027995.1 XM_017027996.1 XM_017027997.1 XM_017027998.1 XM_017027999.1 XM_017028000.1 XM_017028001.1 XM_017028002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4977216 c.544C>A p.Q182K Chr.20: 59984641-59984641 on Build GRCh38
4099918 c.591T>C p.T197T Chr.20: 59984688-59984688 on Build GRCh38
4839687 c.568G>T p.V190F Chr.20: 59984665-59984665 on Build GRCh38
3707893 c.592C>T p.P198S Chr.20: 59984689-59984689 on Build GRCh38
1483805 c.585A>G p.E195E Chr.20: 59984682-59984682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150599622 Chr.20: 59984631-59984631 on Build GRCh38
rs770515588 Chr.20: 59984607-59984607 on Build GRCh38
rs745666352 Chr.20: 59984635-59984635 on Build GRCh38
rs963239002 Chr.20: 59984515-59984515 on Build GRCh38
rs557419371 Chr.20: 59984575-59984575 on Build GRCh38
rs148515614 Chr.20: 59984490-59984490 on Build GRCh38
rs762689603 Chr.20: 59984658-59984658 on Build GRCh38
rs1030349295 Chr.20: 59984472-59984472 on Build GRCh38
rs982871224 Chr.20: 59984469-59984469 on Build GRCh38
rs775177946 Chr.20: 59984614-59984614 on Build GRCh38
rs141839224 Chr.20: 59984609-59984609 on Build GRCh38
rs1041195048 Chr.20: 59984648-59984648 on Build GRCh38
rs760430859 Chr.20: 59984628-59984628 on Build GRCh38
rs1001402526 Chr.20: 59984620-59984620 on Build GRCh38
rs386815753 Chr.20: 59984575-59984577 on Build GRCh38
rs909030108 Chr.20: 59984494-59984494 on Build GRCh38
rs1474909 Chr.20: 59984599-59984599 on Build GRCh38
rs113646744 Chr.20: 59984470-59984470 on Build GRCh38
rs777189094 Chr.20: 59984573-59984573 on Build GRCh38
rs1057173677 Chr.20: 59984576-59984576 on Build GRCh38
rs1043028453 Chr.20: 59984529-59984529 on Build GRCh38
rs952853862 Chr.20: 59984462-59984463 on Build GRCh38
rs186140944 Chr.20: 59984663-59984663 on Build GRCh38
rs764655668 Chr.20: 59984657-59984657 on Build GRCh38
rs918964707 Chr.20: 59984574-59984574 on Build GRCh38
rs771591143 Chr.20: 59984485-59984485 on Build GRCh38
rs780572708 Chr.20: 59984672-59984672 on Build GRCh38
rs758854327 Chr.20: 59984668-59984668 on Build GRCh38
rs578227845 Chr.20: 59984577-59984577 on Build GRCh38
rs763785040 Chr.20: 59984630-59984630 on Build GRCh38
rs557944532 Chr.20: 59984667-59984667 on Build GRCh38

View Full Product Details