Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 100275371-100275632 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLOC1S2
Entrez Gene ID
282991
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 2
Gene Aliases
BLOS2, BORCS2, CEAP, CEAP11
UniGene
Hs.34906
Gene Chromosome Location
Chr.10: 100273278-100286712 on Build GRCh38
Transcripts
NM_001001342.2 NM_001282436.1 NM_001282437.1 NM_001282438.1 NM_001282439.1 NM_173809.4

SNP Detail

SNP ID Chromosome Location
rs769220731 Chr.10: 100275489-100275489 on Build GRCh38
rs372662182 Chr.10: 100275425-100275426 on Build GRCh38
rs528889155 Chr.10: 100275597-100275597 on Build GRCh38
rs199549929 Chr.10: 100275487-100275487 on Build GRCh38
rs750854273 Chr.10: 100275524-100275524 on Build GRCh38
rs940085997 Chr.10: 100275603-100275603 on Build GRCh38
rs764290911 Chr.10: 100275475-100275475 on Build GRCh38
rs200508104 Chr.10: 100275436-100275436 on Build GRCh38
rs751808615 Chr.10: 100275483-100275483 on Build GRCh38
rs756443794 Chr.10: 100275534-100275534 on Build GRCh38
rs765277782 Chr.10: 100275451-100275451 on Build GRCh38
rs757590990 Chr.10: 100275502-100275502 on Build GRCh38
rs191865321 Chr.10: 100275426-100275426 on Build GRCh38
rs200793957 Chr.10: 100275435-100275435 on Build GRCh38
rs76271549 Chr.10: 100275602-100275602 on Build GRCh38
rs760783037 Chr.10: 100275445-100275445 on Build GRCh38
rs759388142 Chr.10: 100275448-100275448 on Build GRCh38
rs775365656 Chr.10: 100275438-100275439 on Build GRCh38
rs760345153 Chr.10: 100275439-100275439 on Build GRCh38
rs917944995 Chr.10: 100275554-100275554 on Build GRCh38
rs372277481 Chr.10: 100275405-100275405 on Build GRCh38
rs202179870 Chr.10: 100275463-100275463 on Build GRCh38
rs747231672 Chr.10: 100275538-100275543 on Build GRCh38
rs373459377 Chr.10: 100275413-100275413 on Build GRCh38
rs186329518 Chr.10: 100275400-100275400 on Build GRCh38
rs781685200 Chr.10: 100275515-100275515 on Build GRCh38
rs917899647 Chr.10: 100275412-100275412 on Build GRCh38
rs375874506 Chr.10: 100275545-100275545 on Build GRCh38
rs774829344 Chr.10: 100275518-100275518 on Build GRCh38
rs781387095 Chr.10: 100275426-100275426 on Build GRCh38
rs769890995 Chr.10: 100275435-100275436 on Build GRCh38
rs763047929 Chr.10: 100275466-100275466 on Build GRCh38
rs766271426 Chr.10: 100275507-100275507 on Build GRCh38

View Full Product Details