Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 148590849-148591120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf33
Entrez Gene ID
202865
Gene Name
chromosome 7 open reading frame 33
Gene Aliases
-
UniGene
Hs.352212
Gene Chromosome Location
Chr.7: 148590565-148615860 on Build GRCh38
Transcripts
NM_145304.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6283240 c.180G>A p.L60L Chr.7: 148591105-148591105 on Build GRCh38
6471744 c.155G>C p.R52T Chr.7: 148591080-148591080 on Build GRCh38
3778206 c.191C>A p.T64K Chr.7: 148591116-148591116 on Build GRCh38
3663013 c.115C>A p.R39S Chr.7: 148591040-148591040 on Build GRCh38
3635725 c.52C>T p.P18S Chr.7: 148590977-148590977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769492751 Chr.7: 148591081-148591081 on Build GRCh38
rs759489205 Chr.7: 148591040-148591040 on Build GRCh38
rs769524388 Chr.7: 148591014-148591014 on Build GRCh38
rs373993336 Chr.7: 148590916-148590916 on Build GRCh38
rs575605385 Chr.7: 148591069-148591069 on Build GRCh38
rs111320068 Chr.7: 148591043-148591043 on Build GRCh38
rs373549941 Chr.7: 148591085-148591085 on Build GRCh38
rs762971919 Chr.7: 148590938-148590938 on Build GRCh38
rs145588023 Chr.7: 148590962-148590962 on Build GRCh38
rs780799842 Chr.7: 148591062-148591062 on Build GRCh38
rs780485421 Chr.7: 148590959-148590959 on Build GRCh38
rs369072198 Chr.7: 148590881-148590881 on Build GRCh38
rs776331085 Chr.7: 148590995-148590995 on Build GRCh38
rs769275398 Chr.7: 148590882-148590882 on Build GRCh38
rs539305324 Chr.7: 148591045-148591045 on Build GRCh38
rs959214895 Chr.7: 148590965-148590965 on Build GRCh38
rs950736224 Chr.7: 148591042-148591042 on Build GRCh38
rs549712040 Chr.7: 148591084-148591084 on Build GRCh38
rs376766533 Chr.7: 148591026-148591026 on Build GRCh38
rs369276406 Chr.7: 148590915-148590915 on Build GRCh38
rs147689956 Chr.7: 148590988-148590988 on Build GRCh38
rs62624492 Chr.7: 148590928-148590928 on Build GRCh38
rs902144031 Chr.7: 148590951-148590951 on Build GRCh38
rs370070335 Chr.7: 148591060-148591060 on Build GRCh38
rs758770856 Chr.7: 148591052-148591052 on Build GRCh38
rs778398385 Chr.7: 148591059-148591059 on Build GRCh38
rs759296225 Chr.7: 148591010-148591010 on Build GRCh38
rs764108415 Chr.7: 148591034-148591034 on Build GRCh38
rs555323349 Chr.7: 148591068-148591068 on Build GRCh38
rs755437640 Chr.7: 148590964-148590964 on Build GRCh38
rs752616222 Chr.7: 148590933-148590933 on Build GRCh38
rs748800881 Chr.7: 148591091-148591091 on Build GRCh38
rs979694947 Chr.7: 148590929-148590929 on Build GRCh38
rs775386548 Chr.7: 148591025-148591025 on Build GRCh38
rs745483086 Chr.7: 148591075-148591075 on Build GRCh38
rs376752947 Chr.7: 148590892-148590892 on Build GRCh38
rs755254079 Chr.7: 148591046-148591046 on Build GRCh38
rs765216372 Chr.7: 148590932-148590932 on Build GRCh38
rs868247963 Chr.7: 148590970-148590970 on Build GRCh38
rs750523525 Chr.7: 148590952-148590952 on Build GRCh38
rs533736703 Chr.7: 148590982-148590982 on Build GRCh38
rs756799041 Chr.7: 148591061-148591061 on Build GRCh38
rs760275756 Chr.7: 148590902-148590902 on Build GRCh38
rs770555915 Chr.7: 148590989-148590989 on Build GRCh38
rs143086119 Chr.7: 148591047-148591047 on Build GRCh38
rs142396932 Chr.7: 148591029-148591029 on Build GRCh38
rs771535280 Chr.7: 148590900-148590900 on Build GRCh38
rs776627436 Chr.7: 148590987-148590987 on Build GRCh38
rs375844560 Chr.7: 148590879-148590879 on Build GRCh38
rs770845401 Chr.7: 148590971-148590971 on Build GRCh38
rs112256370 Chr.7: 148590958-148590958 on Build GRCh38
rs1039258976 Chr.7: 148591080-148591080 on Build GRCh38
rs145680505 Chr.7: 148591048-148591048 on Build GRCh38
rs372006128 Chr.7: 148590887-148590887 on Build GRCh38
rs749685922 Chr.7: 148590878-148590878 on Build GRCh38
rs748551171 Chr.7: 148590966-148590966 on Build GRCh38

View Full Product Details