Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48459954-48460207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRIP
Entrez Gene ID
84126
Gene Name
ATR interacting protein
Gene Aliases
-
UniGene
Hs.694840
Gene Chromosome Location
Chr.3: 48446710-48465655 on Build GRCh38
Transcripts
NM_001271022.1 NM_001271023.1 NM_032166.3 NM_130384.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3916213 c.1112C>T p.S371F Chr.3: 48460166-48460166 on Build GRCh38
1045822 c.1139A>T p.N380I Chr.3: 48460193-48460193 on Build GRCh38
3594914 c.1125G>A p.L375L Chr.3: 48460179-48460179 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375560807 Chr.3: 48460076-48460076 on Build GRCh38
rs139437729 Chr.3: 48460175-48460175 on Build GRCh38
rs1041022900 Chr.3: 48460144-48460144 on Build GRCh38
rs368490195 Chr.3: 48460153-48460153 on Build GRCh38
rs201840650 Chr.3: 48460152-48460152 on Build GRCh38
rs550764790 Chr.3: 48460047-48460047 on Build GRCh38
rs970991529 Chr.3: 48460054-48460054 on Build GRCh38
rs768433947 Chr.3: 48460094-48460094 on Build GRCh38
rs561287676 Chr.3: 48460080-48460081 on Build GRCh38
rs145979024 Chr.3: 48460120-48460120 on Build GRCh38
rs772207358 Chr.3: 48460083-48460083 on Build GRCh38
rs72556557 Chr.3: 48460025-48460025 on Build GRCh38
rs765816483 Chr.3: 48460143-48460143 on Build GRCh38
rs750452581 Chr.3: 48460128-48460128 on Build GRCh38
rs571666281 Chr.3: 48460140-48460140 on Build GRCh38
rs757661922 Chr.3: 48460174-48460174 on Build GRCh38
rs766240937 Chr.3: 48460089-48460092 on Build GRCh38
rs1053507575 Chr.3: 48460110-48460110 on Build GRCh38
rs778152912 Chr.3: 48460155-48460155 on Build GRCh38
rs550209443 Chr.3: 48460127-48460127 on Build GRCh38
rs567664071 Chr.3: 48460106-48460106 on Build GRCh38
rs890361567 Chr.3: 48460135-48460135 on Build GRCh38
rs373800464 Chr.3: 48460187-48460187 on Build GRCh38
rs925406672 Chr.3: 48460166-48460166 on Build GRCh38
rs1050710147 Chr.3: 48460131-48460131 on Build GRCh38
rs760821974 Chr.3: 48460092-48460092 on Build GRCh38
rs775768665 Chr.3: 48460086-48460086 on Build GRCh38
rs754182066 Chr.3: 48460161-48460161 on Build GRCh38
rs981760589 Chr.3: 48460060-48460060 on Build GRCh38
rs959675827 Chr.3: 48460042-48460042 on Build GRCh38
rs745920772 Chr.3: 48460078-48460078 on Build GRCh38
rs771453291 Chr.3: 48460067-48460067 on Build GRCh38
rs139858543 Chr.3: 48460179-48460179 on Build GRCh38
rs530072293 Chr.3: 48460185-48460185 on Build GRCh38

View Full Product Details