Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22180279-22180552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP1
Entrez Gene ID
649
Gene Name
bone morphogenetic protein 1
Gene Aliases
OI13, PCOLC, PCP, PCP2, TLD
UniGene
Hs.1274
Gene Chromosome Location
Chr.8: 22165140-22212326 on Build GRCh38
Transcripts
NM_001199.3 NM_006129.4 NR_033403.1 NR_033404.1 XM_006716386.2 XM_011544617.1 XM_017013738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6611832 c.1044C>T p.C348C Chr.8: 22180450-22180450 on Build GRCh38
4665811 c.962-1G>T p.? Chr.8: 22180367-22180367 on Build GRCh38
4006917 c.982G>C p.D328H Chr.8: 22180388-22180388 on Build GRCh38
1098048 c.1045G>A p.V349M Chr.8: 22180451-22180451 on Build GRCh38
5898938 c.1072G>A p.E358K Chr.8: 22180478-22180478 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037116854 Chr.8: 22180457-22180457 on Build GRCh38
rs758416404 Chr.8: 22180321-22180321 on Build GRCh38
rs775386055 Chr.8: 22180332-22180332 on Build GRCh38
rs767911481 Chr.8: 22180468-22180468 on Build GRCh38
rs771543253 Chr.8: 22180435-22180435 on Build GRCh38
rs755452404 Chr.8: 22180465-22180465 on Build GRCh38
rs750808875 Chr.8: 22180398-22180398 on Build GRCh38
rs760573256 Chr.8: 22180336-22180336 on Build GRCh38
rs768599763 Chr.8: 22180513-22180513 on Build GRCh38
rs368845449 Chr.8: 22180330-22180330 on Build GRCh38
rs745519737 Chr.8: 22180488-22180488 on Build GRCh38
rs932431314 Chr.8: 22180478-22180478 on Build GRCh38
rs778155776 Chr.8: 22180487-22180487 on Build GRCh38
rs146604892 Chr.8: 22180412-22180412 on Build GRCh38
rs781447638 Chr.8: 22180318-22180318 on Build GRCh38
rs531605451 Chr.8: 22180459-22180459 on Build GRCh38
rs770623430 Chr.8: 22180323-22180323 on Build GRCh38
rs866070053 Chr.8: 22180473-22180473 on Build GRCh38
rs78637678 Chr.8: 22180331-22180331 on Build GRCh38
rs966721966 Chr.8: 22180299-22180299 on Build GRCh38
rs763251398 Chr.8: 22180524-22180524 on Build GRCh38
rs776421848 Chr.8: 22180445-22180445 on Build GRCh38
rs140177900 Chr.8: 22180523-22180523 on Build GRCh38
rs778562515 Chr.8: 22180421-22180421 on Build GRCh38
rs774932792 Chr.8: 22180438-22180438 on Build GRCh38
rs143279632 Chr.8: 22180451-22180451 on Build GRCh38
rs371320295 Chr.8: 22180475-22180475 on Build GRCh38
rs377168806 Chr.8: 22180345-22180345 on Build GRCh38
rs373658996 Chr.8: 22180442-22180442 on Build GRCh38
rs766519750 Chr.8: 22180528-22180528 on Build GRCh38
rs1049029535 Chr.8: 22180447-22180447 on Build GRCh38
rs200412717 Chr.8: 22180350-22180350 on Build GRCh38
rs781604387 Chr.8: 22180411-22180411 on Build GRCh38
rs776741766 Chr.8: 22180318-22180318 on Build GRCh38
rs559069167 Chr.8: 22180343-22180343 on Build GRCh38
rs780328194 Chr.8: 22180378-22180378 on Build GRCh38
rs755501101 Chr.8: 22180407-22180407 on Build GRCh38
rs879048776 Chr.8: 22180432-22180432 on Build GRCh38
rs369717073 Chr.8: 22180365-22180365 on Build GRCh38
rs375953306 Chr.8: 22180500-22180500 on Build GRCh38
rs779559164 Chr.8: 22180497-22180497 on Build GRCh38
rs758573319 Chr.8: 22180369-22180369 on Build GRCh38
rs745571172 Chr.8: 22180433-22180433 on Build GRCh38
rs752073136 Chr.8: 22180390-22180390 on Build GRCh38
rs746954049 Chr.8: 22180439-22180439 on Build GRCh38
rs750489501 Chr.8: 22180364-22180364 on Build GRCh38
rs776455120 Chr.8: 22180515-22180515 on Build GRCh38
rs763947051 Chr.8: 22180340-22180340 on Build GRCh38
rs758105387 Chr.8: 22180493-22180493 on Build GRCh38
rs199556894 Chr.8: 22180474-22180474 on Build GRCh38
rs577420762 Chr.8: 22180458-22180458 on Build GRCh38
rs770500738 Chr.8: 22180413-22180413 on Build GRCh38
rs149174555 Chr.8: 22180450-22180450 on Build GRCh38

View Full Product Details