Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 79294021-79294282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD34C
Entrez Gene ID
390616
Gene Name
ankyrin repeat domain 34C
Gene Aliases
-
UniGene
Hs.531231
Gene Chromosome Location
Chr.15: 79282804-79298239 on Build GRCh38
Transcripts
NM_001146341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1563338 c.798G>A p.S266S Chr.15: 79294082-79294082 on Build GRCh38
965471 c.803G>A p.R268H Chr.15: 79294087-79294087 on Build GRCh38
5526675 c.764G>A p.W255* Chr.15: 79294048-79294048 on Build GRCh38
404680 c.747G>T p.R249S Chr.15: 79294031-79294031 on Build GRCh38
6443476 c.851G>T p.S284I Chr.15: 79294135-79294135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971080019 Chr.15: 79294074-79294074 on Build GRCh38
rs1021099513 Chr.15: 79294190-79294190 on Build GRCh38
rs759840006 Chr.15: 79294122-79294122 on Build GRCh38
rs927055082 Chr.15: 79294086-79294086 on Build GRCh38
rs1048265317 Chr.15: 79294084-79294084 on Build GRCh38
rs765182814 Chr.15: 79294141-79294141 on Build GRCh38
rs996848300 Chr.15: 79294137-79294137 on Build GRCh38
rs991849623 Chr.15: 79294067-79294067 on Build GRCh38
rs886911869 Chr.15: 79294087-79294087 on Build GRCh38
rs529256936 Chr.15: 79294061-79294061 on Build GRCh38
rs758230110 Chr.15: 79294163-79294163 on Build GRCh38
rs894071657 Chr.15: 79294169-79294169 on Build GRCh38
rs1039697550 Chr.15: 79294184-79294184 on Build GRCh38
rs977604822 Chr.15: 79294233-79294233 on Build GRCh38
rs1025712446 Chr.15: 79294259-79294259 on Build GRCh38
rs777603919 Chr.15: 79294201-79294201 on Build GRCh38
rs751790086 Chr.15: 79294261-79294261 on Build GRCh38
rs560579919 Chr.15: 79294060-79294060 on Build GRCh38
rs992847450 Chr.15: 79294063-79294063 on Build GRCh38
rs763799566 Chr.15: 79294174-79294174 on Build GRCh38
rs940117303 Chr.15: 79294118-79294118 on Build GRCh38
rs753382968 Chr.15: 79294081-79294081 on Build GRCh38
rs147308214 Chr.15: 79294156-79294156 on Build GRCh38
rs758979450 Chr.15: 79294121-79294121 on Build GRCh38
rs931179221 Chr.15: 79294082-79294082 on Build GRCh38
rs752592824 Chr.15: 79294162-79294162 on Build GRCh38
rs1054315690 Chr.15: 79294098-79294098 on Build GRCh38

View Full Product Details