Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 110907531-110907784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBIP1
Entrez Gene ID
92482
Gene Name
BBSome interacting protein 1
Gene Aliases
BBIP10, BBS18, NCRNA00081, bA348N5.3
UniGene
Hs.232543
Gene Chromosome Location
Chr.10: 110898730-110919366 on Build GRCh38
Transcripts
NM_001195304.1 NM_001195305.1 NM_001195306.1 NM_001195307.1 NM_001243783.1

SNP Detail

SNP ID Chromosome Location
rs774118437 Chr.10: 110907641-110907641 on Build GRCh38
rs148407939 Chr.10: 110907640-110907640 on Build GRCh38
rs1023271863 Chr.10: 110907705-110907705 on Build GRCh38
rs766297227 Chr.10: 110907572-110907572 on Build GRCh38
rs565821309 Chr.10: 110907599-110907599 on Build GRCh38
rs529871959 Chr.10: 110907601-110907601 on Build GRCh38
rs535978583 Chr.10: 110907622-110907625 on Build GRCh38
rs759215697 Chr.10: 110907703-110907703 on Build GRCh38
rs927884873 Chr.10: 110907752-110907752 on Build GRCh38
rs776153373 Chr.10: 110907588-110907588 on Build GRCh38
rs537179829 Chr.10: 110907655-110907655 on Build GRCh38
rs751334242 Chr.10: 110907682-110907682 on Build GRCh38
rs117419791 Chr.10: 110907748-110907748 on Build GRCh38
rs569801734 Chr.10: 110907651-110907651 on Build GRCh38
rs1052351751 Chr.10: 110907665-110907665 on Build GRCh38
rs35793075 Chr.10: 110907566-110907566 on Build GRCh38
rs760064445 Chr.10: 110907683-110907683 on Build GRCh38
rs753378493 Chr.10: 110907702-110907702 on Build GRCh38
rs1007112104 Chr.10: 110907607-110907607 on Build GRCh38
rs765922419 Chr.10: 110907689-110907689 on Build GRCh38
rs891137947 Chr.10: 110907680-110907683 on Build GRCh38
rs771976595 Chr.10: 110907669-110907669 on Build GRCh38
rs866047769 Chr.10: 110907749-110907749 on Build GRCh38
rs904098021 Chr.10: 110907653-110907653 on Build GRCh38
rs984212075 Chr.10: 110907697-110907697 on Build GRCh38
rs901302275 Chr.10: 110907643-110907643 on Build GRCh38

View Full Product Details