Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154349504-154349757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLNA
Entrez Gene ID
2316
Gene Name
filamin A
Gene Aliases
ABP-280, ABPX, CSBS, CVD1, FLN, FLN-A, FLN1, FMD, MNS, NHBP, OPD, OPD1, OPD2, XLVD, XMVD
UniGene
Hs.195464
Gene Chromosome Location
Chr.X: 154348529-154374638 on Build GRCh38
Transcripts
NM_001110556.1 NM_001456.3 XM_011531127.1 XM_011531128.1 XM_011531129.1 XM_011531130.1 XM_011531131.1

SNP Detail

SNP ID Chromosome Location
rs782276654 Chr.X: 154349697-154349697 on Build GRCh38
rs782502145 Chr.X: 154349689-154349689 on Build GRCh38
rs781941043 Chr.X: 154349584-154349584 on Build GRCh38
rs782335342 Chr.X: 154349555-154349555 on Build GRCh38
rs372491500 Chr.X: 154349608-154349608 on Build GRCh38
rs781909155 Chr.X: 154349656-154349656 on Build GRCh38
rs782013219 Chr.X: 154349571-154349571 on Build GRCh38
rs782084663 Chr.X: 154349606-154349606 on Build GRCh38
rs375433852 Chr.X: 154349609-154349609 on Build GRCh38
rs782431987 Chr.X: 154349732-154349732 on Build GRCh38
rs782383611 Chr.X: 154349563-154349563 on Build GRCh38
rs112682871 Chr.X: 154349583-154349583 on Build GRCh38
rs187774579 Chr.X: 154349554-154349554 on Build GRCh38
rs782790817 Chr.X: 154349617-154349617 on Build GRCh38
rs782156724 Chr.X: 154349575-154349575 on Build GRCh38
rs909387820 Chr.X: 154349738-154349738 on Build GRCh38
rs782563616 Chr.X: 154349733-154349733 on Build GRCh38
rs201883808 Chr.X: 154349534-154349534 on Build GRCh38
rs148571693 Chr.X: 154349613-154349613 on Build GRCh38
rs782244918 Chr.X: 154349560-154349560 on Build GRCh38
rs782442409 Chr.X: 154349657-154349657 on Build GRCh38
rs985402675 Chr.X: 154349713-154349713 on Build GRCh38
rs782555930 Chr.X: 154349550-154349550 on Build GRCh38
rs782602947 Chr.X: 154349671-154349671 on Build GRCh38
rs200195310 Chr.X: 154349695-154349695 on Build GRCh38
rs971390304 Chr.X: 154349682-154349682 on Build GRCh38
rs981689322 Chr.X: 154349692-154349692 on Build GRCh38
rs1022884327 Chr.X: 154349661-154349661 on Build GRCh38
rs398123623 Chr.X: 154349643-154349650 on Build GRCh38
rs863223626 Chr.X: 154349681-154349681 on Build GRCh38
rs781959191 Chr.X: 154349559-154349559 on Build GRCh38
rs782611801 Chr.X: 154349675-154349675 on Build GRCh38

View Full Product Details