Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29798519-29798780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF22
Entrez Gene ID
3835
Gene Name
kinesin family member 22
Gene Aliases
A-328A3.2, KID, KNSL4, OBP, OBP-1, OBP-2, SEMDJL2
UniGene
Hs.612151
Gene Chromosome Location
Chr.16: 29790062-29805385 on Build GRCh38
Transcripts
NM_001256269.1 NM_001256270.1 NM_007317.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
312341 c.454A>G p.M152V Chr.16: 29798652-29798652 on Build GRCh38
5923154 c.395-4C>T p.? Chr.16: 29798589-29798589 on Build GRCh38
5923155 c.395-3C>T p.? Chr.16: 29798590-29798590 on Build GRCh38
6695204 c.418A>C p.S140R Chr.16: 29798616-29798616 on Build GRCh38
5906027 c.508C>T p.L170F Chr.16: 29798706-29798706 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780641872 Chr.16: 29798722-29798722 on Build GRCh38
rs749613241 Chr.16: 29798586-29798586 on Build GRCh38
rs375396484 Chr.16: 29798758-29798758 on Build GRCh38
rs757801040 Chr.16: 29798567-29798568 on Build GRCh38
rs149549504 Chr.16: 29798643-29798643 on Build GRCh38
rs772828204 Chr.16: 29798760-29798760 on Build GRCh38
rs926677595 Chr.16: 29798696-29798696 on Build GRCh38
rs758780979 Chr.16: 29798719-29798719 on Build GRCh38
rs748373911 Chr.16: 29798757-29798757 on Build GRCh38
rs754495075 Chr.16: 29798552-29798552 on Build GRCh38
rs754532607 Chr.16: 29798553-29798553 on Build GRCh38
rs11545429 Chr.16: 29798727-29798727 on Build GRCh38
rs779428738 Chr.16: 29798546-29798546 on Build GRCh38
rs750783800 Chr.16: 29798586-29798586 on Build GRCh38
rs193922921 Chr.16: 29798641-29798641 on Build GRCh38
rs917054085 Chr.16: 29798651-29798651 on Build GRCh38
rs771255035 Chr.16: 29798615-29798615 on Build GRCh38
rs762171792 Chr.16: 29798660-29798660 on Build GRCh38
rs755824124 Chr.16: 29798699-29798699 on Build GRCh38
rs765108503 Chr.16: 29798662-29798662 on Build GRCh38
rs193922920 Chr.16: 29798640-29798640 on Build GRCh38
rs371843824 Chr.16: 29798678-29798678 on Build GRCh38
rs779247770 Chr.16: 29798700-29798700 on Build GRCh38
rs758898747 Chr.16: 29798636-29798681 on Build GRCh38
rs374331057 Chr.16: 29798755-29798755 on Build GRCh38
rs765047195 Chr.16: 29798604-29798604 on Build GRCh38
rs780849125 Chr.16: 29798558-29798558 on Build GRCh38
rs779557868 Chr.16: 29798743-29798745 on Build GRCh38
rs775372716 Chr.16: 29798642-29798642 on Build GRCh38
rs750989417 Chr.16: 29798687-29798687 on Build GRCh38
rs867074945 Chr.16: 29798625-29798625 on Build GRCh38
rs776958584 Chr.16: 29798647-29798647 on Build GRCh38
rs766356873 Chr.16: 29798685-29798685 on Build GRCh38
rs368750627 Chr.16: 29798632-29798632 on Build GRCh38
rs193922922 Chr.16: 29798644-29798644 on Build GRCh38
rs750307687 Chr.16: 29798667-29798667 on Build GRCh38
rs765869790 Chr.16: 29798585-29798585 on Build GRCh38
rs752413453 Chr.16: 29798695-29798695 on Build GRCh38
rs767054899 Chr.16: 29798694-29798694 on Build GRCh38
rs772635300 Chr.16: 29798638-29798638 on Build GRCh38
rs746349728 Chr.16: 29798634-29798634 on Build GRCh38
rs758077486 Chr.16: 29798705-29798705 on Build GRCh38
rs1056247785 Chr.16: 29798732-29798732 on Build GRCh38
rs768829846 Chr.16: 29798748-29798748 on Build GRCh38
rs200923972 Chr.16: 29798688-29798688 on Build GRCh38

View Full Product Details