Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4335086-4335317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIS2
Entrez Gene ID
84662
Gene Name
GLIS family zinc finger 2
Gene Aliases
NKL, NPHP7
UniGene
Hs.592087
Gene Chromosome Location
Chr.16: 4314761-4339597 on Build GRCh38
Transcripts
NM_001318918.1 NM_032575.2 XM_005255641.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6144591 c.589G>A p.E197K Chr.16: 4335126-4335126 on Build GRCh38
7190068 c.599C>T p.A200V Chr.16: 4335136-4335136 on Build GRCh38
2920340 c.690_692delCAA p.N231delN Chr.16: 4335308-4335310 on Build GRCh38
703772 c.694G>T p.E232* Chr.16: 4335312-4335312 on Build GRCh38
4687191 c.632G>A p.R211H Chr.16: 4335169-4335169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043516792 Chr.16: 4335126-4335126 on Build GRCh38
rs1053391302 Chr.16: 4335177-4335177 on Build GRCh38
rs759708250 Chr.16: 4335199-4335199 on Build GRCh38
rs780572049 Chr.16: 4335231-4335231 on Build GRCh38
rs973774997 Chr.16: 4335138-4335138 on Build GRCh38
rs758017634 Chr.16: 4335217-4335217 on Build GRCh38
rs942962442 Chr.16: 4335242-4335242 on Build GRCh38
rs758072621 Chr.16: 4335152-4335152 on Build GRCh38
rs763992882 Chr.16: 4335283-4335283 on Build GRCh38
rs764396971 Chr.16: 4335214-4335214 on Build GRCh38
rs1014571686 Chr.16: 4335179-4335179 on Build GRCh38
rs749133207 Chr.16: 4335256-4335256 on Build GRCh38
rs768457255 Chr.16: 4335165-4335165 on Build GRCh38
rs761135861 Chr.16: 4335298-4335298 on Build GRCh38
rs748040914 Chr.16: 4335240-4335240 on Build GRCh38
rs150608208 Chr.16: 4335220-4335220 on Build GRCh38
rs936293701 Chr.16: 4335153-4335153 on Build GRCh38
rs756613792 Chr.16: 4335213-4335213 on Build GRCh38
rs745469791 Chr.16: 4335151-4335151 on Build GRCh38
rs377111318 Chr.16: 4335178-4335178 on Build GRCh38
rs771034747 Chr.16: 4335257-4335257 on Build GRCh38
rs769873681 Chr.16: 4335245-4335245 on Build GRCh38
rs746571582 Chr.16: 4335164-4335164 on Build GRCh38
rs774262912 Chr.16: 4335196-4335196 on Build GRCh38
rs372267495 Chr.16: 4335173-4335173 on Build GRCh38
rs763272271 Chr.16: 4335116-4335116 on Build GRCh38
rs767562840 Chr.16: 4335281-4335281 on Build GRCh38
rs776183248 Chr.16: 4335127-4335129 on Build GRCh38
rs761100838 Chr.16: 4335205-4335206 on Build GRCh38
rs746488896 Chr.16: 4335224-4335224 on Build GRCh38
rs751681231 Chr.16: 4335129-4335129 on Build GRCh38
rs947449559 Chr.16: 4335105-4335105 on Build GRCh38
rs752979914 Chr.16: 4335143-4335143 on Build GRCh38
rs200062704 Chr.16: 4335258-4335258 on Build GRCh38
rs754235272 Chr.16: 4335216-4335216 on Build GRCh38
rs764577088 Chr.16: 4335227-4335228 on Build GRCh38
rs754584715 Chr.16: 4335228-4335228 on Build GRCh38
rs750742689 Chr.16: 4335107-4335107 on Build GRCh38
rs775835280 Chr.16: 4335249-4335249 on Build GRCh38
rs767894873 Chr.16: 4335203-4335203 on Build GRCh38
rs369868465 Chr.16: 4335215-4335215 on Build GRCh38
rs766508781 Chr.16: 4335125-4335125 on Build GRCh38
rs776689106 Chr.16: 4335166-4335166 on Build GRCh38
rs140609747 Chr.16: 4335188-4335188 on Build GRCh38
rs756525846 Chr.16: 4335145-4335145 on Build GRCh38
rs753309810 Chr.16: 4335209-4335209 on Build GRCh38
rs775458398 Chr.16: 4335288-4335288 on Build GRCh38
rs778528093 Chr.16: 4335150-4335150 on Build GRCh38
rs773154166 Chr.16: 4335180-4335180 on Build GRCh38
rs771595358 Chr.16: 4335181-4335181 on Build GRCh38
rs769944123 Chr.16: 4335219-4335219 on Build GRCh38
rs368985225 Chr.16: 4335293-4335293 on Build GRCh38
rs372248083 Chr.16: 4335250-4335250 on Build GRCh38
rs186957679 Chr.16: 4335137-4335137 on Build GRCh38
rs572594716 Chr.16: 4335261-4335261 on Build GRCh38
rs201509639 Chr.16: 4335253-4335253 on Build GRCh38
rs1011610705 Chr.16: 4335259-4335259 on Build GRCh38

View Full Product Details