Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 129455167-129455440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STRIP2
Entrez Gene ID
57464
Gene Name
striatin interacting protein 2
Gene Aliases
FAM40B, FAR11B
UniGene
Hs.489988
Gene Chromosome Location
Chr.7: 129434433-129488399 on Build GRCh38
Transcripts
NM_001134336.1 NM_020704.2 XM_006716069.3 XM_011516432.2 XM_011516435.1 XM_017012470.1 XM_017012471.1 XM_017012472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
744575 c.776G>T p.G259V Chr.7: 129455313-129455313 on Build GRCh38
6033770 c.817C>T p.L273F Chr.7: 129455354-129455354 on Build GRCh38
7236295 c.755T>C p.M252T Chr.7: 129455292-129455292 on Build GRCh38
3922684 c.792C>T p.F264F Chr.7: 129455329-129455329 on Build GRCh38
390673 c.775G>T p.G259C Chr.7: 129455312-129455312 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936010480 Chr.7: 129455219-129455219 on Build GRCh38
rs762582355 Chr.7: 129455372-129455375 on Build GRCh38
rs886117309 Chr.7: 129455400-129455400 on Build GRCh38
rs769156267 Chr.7: 129455259-129455260 on Build GRCh38
rs142679514 Chr.7: 129455353-129455353 on Build GRCh38
rs747963022 Chr.7: 129455338-129455338 on Build GRCh38
rs1026636030 Chr.7: 129455280-129455280 on Build GRCh38
rs1038773275 Chr.7: 129455324-129455324 on Build GRCh38
rs753527228 Chr.7: 129455395-129455395 on Build GRCh38
rs946279115 Chr.7: 129455238-129455238 on Build GRCh38
rs754443759 Chr.7: 129455230-129455230 on Build GRCh38
rs532922480 Chr.7: 129455235-129455235 on Build GRCh38
rs913237983 Chr.7: 129455229-129455229 on Build GRCh38
rs766050432 Chr.7: 129455379-129455379 on Build GRCh38
rs763439477 Chr.7: 129455335-129455335 on Build GRCh38
rs777491329 Chr.7: 129455260-129455260 on Build GRCh38
rs775627580 Chr.7: 129455333-129455333 on Build GRCh38
rs951367894 Chr.7: 129455305-129455305 on Build GRCh38
rs900215603 Chr.7: 129455331-129455331 on Build GRCh38
rs1017993964 Chr.7: 129455225-129455225 on Build GRCh38
rs775115772 Chr.7: 129455348-129455349 on Build GRCh38
rs755768842 Chr.7: 129455211-129455211 on Build GRCh38
rs764595855 Chr.7: 129455345-129455345 on Build GRCh38
rs373210698 Chr.7: 129455208-129455208 on Build GRCh38
rs370579791 Chr.7: 129455278-129455278 on Build GRCh38
rs778811426 Chr.7: 129455319-129455319 on Build GRCh38
rs1004788339 Chr.7: 129455406-129455406 on Build GRCh38
rs150002869 Chr.7: 129455297-129455297 on Build GRCh38
rs746730562 Chr.7: 129455264-129455264 on Build GRCh38
rs959906983 Chr.7: 129455409-129455409 on Build GRCh38
rs748083251 Chr.7: 129455250-129455250 on Build GRCh38
rs754601361 Chr.7: 129455415-129455415 on Build GRCh38
rs189583337 Chr.7: 129455322-129455322 on Build GRCh38
rs778453356 Chr.7: 129455232-129455232 on Build GRCh38
rs762115684 Chr.7: 129455375-129455375 on Build GRCh38
rs199882915 Chr.7: 129455252-129455252 on Build GRCh38
rs779625454 Chr.7: 129455212-129455212 on Build GRCh38
rs771305258 Chr.7: 129455267-129455267 on Build GRCh38
rs201924047 Chr.7: 129455228-129455228 on Build GRCh38
rs752486134 Chr.7: 129455358-129455358 on Build GRCh38
rs891281874 Chr.7: 129455314-129455314 on Build GRCh38

View Full Product Details