Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12924793-12925018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FARSA
Entrez Gene ID
2193
Gene Name
phenylalanyl-tRNA synthetase alpha subunit
Gene Aliases
CML33, FARSL, FARSLA, FRSA, PheHA
UniGene
Hs.23111
Gene Chromosome Location
Chr.19: 12922470-12933744 on Build GRCh38
Transcripts
NM_004461.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6660621 c.952G>A p.E318K Chr.19: 12924978-12924978 on Build GRCh38
5388693 c.992C>T p.A331V Chr.19: 12924938-12924938 on Build GRCh38
5056248 c.1000C>T p.R334C Chr.19: 12924930-12924930 on Build GRCh38
6305792 c.951C>A p.D317E Chr.19: 12924979-12924979 on Build GRCh38
991541 c.1029G>A p.K343K Chr.19: 12924805-12924805 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754958871 Chr.19: 12924827-12924827 on Build GRCh38
rs766719157 Chr.19: 12924947-12924947 on Build GRCh38
rs778570693 Chr.19: 12924981-12924981 on Build GRCh38
rs777614456 Chr.19: 12924900-12924900 on Build GRCh38
rs770576117 Chr.19: 12924912-12924912 on Build GRCh38
rs750617542 Chr.19: 12924822-12924822 on Build GRCh38
rs933823166 Chr.19: 12924964-12924964 on Build GRCh38
rs202082142 Chr.19: 12924886-12924886 on Build GRCh38
rs757893905 Chr.19: 12924985-12924985 on Build GRCh38
rs368444217 Chr.19: 12924869-12924869 on Build GRCh38
rs751690404 Chr.19: 12924887-12924887 on Build GRCh38
rs543097283 Chr.19: 12924882-12924882 on Build GRCh38
rs200279751 Chr.19: 12924961-12924961 on Build GRCh38
rs964440017 Chr.19: 12924930-12924930 on Build GRCh38
rs200214203 Chr.19: 12924837-12924837 on Build GRCh38
rs750660467 Chr.19: 12924883-12924883 on Build GRCh38
rs777334223 Chr.19: 12924850-12924850 on Build GRCh38
rs771568804 Chr.19: 12924919-12924919 on Build GRCh38
rs370272213 Chr.19: 12924889-12924889 on Build GRCh38
rs772716716 Chr.19: 12924888-12924888 on Build GRCh38
rs773702821 Chr.19: 12924929-12924929 on Build GRCh38
rs752638663 Chr.19: 12924972-12924972 on Build GRCh38
rs371864272 Chr.19: 12924891-12924891 on Build GRCh38
rs770490430 Chr.19: 12924860-12924860 on Build GRCh38
rs560812068 Chr.19: 12924823-12924823 on Build GRCh38
rs368255552 Chr.19: 12924816-12924816 on Build GRCh38
rs753647514 Chr.19: 12924978-12924978 on Build GRCh38
rs868407433 Chr.19: 12924957-12924957 on Build GRCh38
rs767616044 Chr.19: 12924968-12924968 on Build GRCh38
rs531815311 Chr.19: 12924866-12924866 on Build GRCh38
rs8107173 Chr.19: 12924824-12924824 on Build GRCh38
rs773847239 Chr.19: 12924861-12924861 on Build GRCh38
rs35087277 Chr.19: 12924908-12924908 on Build GRCh38
rs747372521 Chr.19: 12924870-12924870 on Build GRCh38
rs113324767 Chr.19: 12924902-12924902 on Build GRCh38
rs537993454 Chr.19: 12924835-12924835 on Build GRCh38
rs778480926 Chr.19: 12924917-12924917 on Build GRCh38
rs532102742 Chr.19: 12924899-12924899 on Build GRCh38
rs777505492 Chr.19: 12924975-12924975 on Build GRCh38
rs975525475 Chr.19: 12924956-12924956 on Build GRCh38
rs770114701 Chr.19: 12924926-12924926 on Build GRCh38
rs748920650 Chr.19: 12924854-12924854 on Build GRCh38
rs751780205 Chr.19: 12924959-12924959 on Build GRCh38
rs201276620 Chr.19: 12924918-12924918 on Build GRCh38
rs192239572 Chr.19: 12924841-12924841 on Build GRCh38
rs755048856 Chr.19: 12924892-12924892 on Build GRCh38
rs762114185 Chr.19: 12924881-12924881 on Build GRCh38
rs759933679 Chr.19: 12924831-12924831 on Build GRCh38
rs114790582 Chr.19: 12924979-12924979 on Build GRCh38
rs757585686 Chr.19: 12924814-12924814 on Build GRCh38
rs763339832 Chr.19: 12924934-12924934 on Build GRCh38
rs142135169 Chr.19: 12924983-12924983 on Build GRCh38
rs1024190533 Chr.19: 12924879-12924879 on Build GRCh38
rs769594954 Chr.19: 12924842-12924842 on Build GRCh38
rs762358431 Chr.19: 12924925-12924925 on Build GRCh38

View Full Product Details