Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 78927033-78927258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRL4
Entrez Gene ID
64123
Gene Name
adhesion G protein-coupled receptor L4
Gene Aliases
ELTD1, ETL, KPG_003
UniGene
Hs.132314
Gene Chromosome Location
Chr.1: 78889764-79006810 on Build GRCh38
Transcripts
NM_022159.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2242949 c.878-2A>G p.? Chr.1: 78927093-78927093 on Build GRCh38
260673 c.920C>A p.P307H Chr.1: 78927049-78927049 on Build GRCh38
7237826 c.899T>C p.V300A Chr.1: 78927070-78927070 on Build GRCh38
4010074 c.929C>T p.S310L Chr.1: 78927040-78927040 on Build GRCh38
912427 c.911G>T p.S304I Chr.1: 78927058-78927058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756308867 Chr.1: 78927078-78927079 on Build GRCh38
rs777361488 Chr.1: 78927149-78927149 on Build GRCh38
rs538159822 Chr.1: 78927219-78927219 on Build GRCh38
rs571169380 Chr.1: 78927209-78927209 on Build GRCh38
rs566704899 Chr.1: 78927147-78927147 on Build GRCh38
rs762367429 Chr.1: 78927233-78927233 on Build GRCh38
rs17412855 Chr.1: 78927136-78927136 on Build GRCh38
rs754510379 Chr.1: 78927069-78927069 on Build GRCh38
rs771199505 Chr.1: 78927105-78927105 on Build GRCh38
rs534158819 Chr.1: 78927179-78927179 on Build GRCh38
rs748635862 Chr.1: 78927076-78927076 on Build GRCh38
rs776067216 Chr.1: 78927203-78927203 on Build GRCh38
rs772334659 Chr.1: 78927081-78927081 on Build GRCh38
rs747328995 Chr.1: 78927096-78927096 on Build GRCh38
rs944992050 Chr.1: 78927200-78927200 on Build GRCh38
rs181298847 Chr.1: 78927160-78927160 on Build GRCh38
rs777989559 Chr.1: 78927140-78927143 on Build GRCh38
rs776811969 Chr.1: 78927124-78927124 on Build GRCh38
rs752731521 Chr.1: 78927062-78927066 on Build GRCh38
rs12754818 Chr.1: 78927071-78927071 on Build GRCh38
rs779841411 Chr.1: 78927066-78927066 on Build GRCh38
rs376746737 Chr.1: 78927062-78927062 on Build GRCh38
rs184032174 Chr.1: 78927187-78927187 on Build GRCh38
rs549448481 Chr.1: 78927225-78927225 on Build GRCh38
rs752680093 Chr.1: 78927088-78927088 on Build GRCh38
rs567975792 Chr.1: 78927135-78927135 on Build GRCh38
rs773556383 Chr.1: 78927086-78927086 on Build GRCh38
rs748860340 Chr.1: 78927067-78927067 on Build GRCh38
rs372682441 Chr.1: 78927142-78927142 on Build GRCh38
rs549701367 Chr.1: 78927132-78927132 on Build GRCh38

View Full Product Details