Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22098226-22098457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM160B2
Entrez Gene ID
64760
Gene Name
family with sequence similarity 160 member B2
Gene Aliases
RAI16
UniGene
Hs.491223
Gene Chromosome Location
Chr.8: 22089150-22104380 on Build GRCh38
Transcripts
NM_022749.5 XM_005273602.3 XM_006716381.2 XM_006716385.3 XM_017013732.1 XM_017013733.1 XM_017013734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5011886 c.769A>G p.K257E Chr.8: 22098423-22098423 on Build GRCh38
1097992 c.724G>C p.E242Q Chr.8: 22098266-22098266 on Build GRCh38
5184366 c.700G>A p.E234K Chr.8: 22098242-22098242 on Build GRCh38
1743298 c.712G>A p.G238S Chr.8: 22098254-22098254 on Build GRCh38
6850146 c.773G>A p.S258N Chr.8: 22098427-22098427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028441106 Chr.8: 22098279-22098279 on Build GRCh38
rs772236485 Chr.8: 22098298-22098298 on Build GRCh38
rs769014264 Chr.8: 22098318-22098318 on Build GRCh38
rs760569288 Chr.8: 22098316-22098316 on Build GRCh38
rs759424977 Chr.8: 22098361-22098361 on Build GRCh38
rs762336631 Chr.8: 22098338-22098338 on Build GRCh38
rs770303251 Chr.8: 22098343-22098343 on Build GRCh38
rs200751092 Chr.8: 22098430-22098430 on Build GRCh38
rs778894231 Chr.8: 22098277-22098277 on Build GRCh38
rs752489254 Chr.8: 22098255-22098255 on Build GRCh38
rs188321970 Chr.8: 22098368-22098368 on Build GRCh38
rs757302325 Chr.8: 22098398-22098398 on Build GRCh38
rs955748037 Chr.8: 22098340-22098340 on Build GRCh38
rs144079413 Chr.8: 22098273-22098273 on Build GRCh38
rs780310659 Chr.8: 22098310-22098310 on Build GRCh38
rs146497282 Chr.8: 22098359-22098359 on Build GRCh38
rs779934363 Chr.8: 22098410-22098410 on Build GRCh38
rs372814624 Chr.8: 22098253-22098253 on Build GRCh38
rs767138257 Chr.8: 22098376-22098376 on Build GRCh38
rs1000409059 Chr.8: 22098300-22098300 on Build GRCh38
rs920427856 Chr.8: 22098371-22098371 on Build GRCh38
rs776924995 Chr.8: 22098334-22098334 on Build GRCh38
rs745933677 Chr.8: 22098283-22098283 on Build GRCh38
rs781574018 Chr.8: 22098436-22098436 on Build GRCh38
rs139434422 Chr.8: 22098254-22098254 on Build GRCh38
rs1041153040 Chr.8: 22098257-22098257 on Build GRCh38
rs376449220 Chr.8: 22098307-22098307 on Build GRCh38
rs773782617 Chr.8: 22098345-22098345 on Build GRCh38
rs765894932 Chr.8: 22098251-22098251 on Build GRCh38
rs777641408 Chr.8: 22098262-22098262 on Build GRCh38
rs755965274 Chr.8: 22098259-22098259 on Build GRCh38
rs764069825 Chr.8: 22098386-22098386 on Build GRCh38
rs140882816 Chr.8: 22098434-22098434 on Build GRCh38
rs201438088 Chr.8: 22098408-22098408 on Build GRCh38
rs369587302 Chr.8: 22098267-22098267 on Build GRCh38
rs747442134 Chr.8: 22098317-22098317 on Build GRCh38
rs752395868 Chr.8: 22098380-22098380 on Build GRCh38
rs760343873 Chr.8: 22098385-22098385 on Build GRCh38
rs77609062 Chr.8: 22098383-22098383 on Build GRCh38
rs753837133 Chr.8: 22098270-22098270 on Build GRCh38
rs778888631 Chr.8: 22098406-22098406 on Build GRCh38
rs968982793 Chr.8: 22098287-22098287 on Build GRCh38
rs971191297 Chr.8: 22098367-22098367 on Build GRCh38
rs373199574 Chr.8: 22098362-22098362 on Build GRCh38
rs750530831 Chr.8: 22098407-22098407 on Build GRCh38
rs753648239 Chr.8: 22098393-22098393 on Build GRCh38
rs1047638532 Chr.8: 22098250-22098250 on Build GRCh38

View Full Product Details