Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40767287-40767512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC17
Entrez Gene ID
55192
Gene Name
DnaJ heat shock protein family (Hsp40) member C17
Gene Aliases
-
UniGene
Hs.511069
Gene Chromosome Location
Chr.15: 40767448-40807478 on Build GRCh38
Transcripts
XM_017022401.1 NM_005258.2

Gene Symbol
GCHFR
Entrez Gene ID
2644
Gene Name
GTP cyclohydrolase I feedback regulator
Gene Aliases
GFRP, HsT16933, P35
UniGene
Hs.631717
Gene Chromosome Location
Chr.15: 40764087-40767713 on Build GRCh38
Transcripts
XM_017022401.1 NM_005258.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5016466 c.206_207delGC p.S69fs*>16 Chr.15: 40767300-40767301 on Build GRCh38
4686618 c.213delG p.V73fs*>12 Chr.15: 40767307-40767307 on Build GRCh38
2269809 c.234G>A p.V78V Chr.15: 40767328-40767328 on Build GRCh38
961297 c.222C>T p.G74G Chr.15: 40767316-40767316 on Build GRCh38
6669949 c.228G>A p.T76T Chr.15: 40767322-40767322 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376132308 Chr.15: 40767357-40767357 on Build GRCh38
rs146242086 Chr.15: 40767306-40767306 on Build GRCh38
rs922274748 Chr.15: 40767403-40767403 on Build GRCh38
rs1016812849 Chr.15: 40767479-40767479 on Build GRCh38
rs901965817 Chr.15: 40767430-40767430 on Build GRCh38
rs139646493 Chr.15: 40767346-40767346 on Build GRCh38
rs78081362 Chr.15: 40767456-40767456 on Build GRCh38
rs998012862 Chr.15: 40767435-40767435 on Build GRCh38
rs754646233 Chr.15: 40767375-40767375 on Build GRCh38
rs201305860 Chr.15: 40767398-40767398 on Build GRCh38
rs752258258 Chr.15: 40767383-40767383 on Build GRCh38
rs772366096 Chr.15: 40767323-40767323 on Build GRCh38
rs772079236 Chr.15: 40767457-40767457 on Build GRCh38
rs770814286 Chr.15: 40767313-40767313 on Build GRCh38
rs202144307 Chr.15: 40767332-40767332 on Build GRCh38
rs867123043 Chr.15: 40767387-40767387 on Build GRCh38
rs1008133201 Chr.15: 40767463-40767463 on Build GRCh38
rs754417572 Chr.15: 40767351-40767351 on Build GRCh38
rs759847227 Chr.15: 40767321-40767321 on Build GRCh38
rs745863947 Chr.15: 40767328-40767330 on Build GRCh38
rs768771246 Chr.15: 40767351-40767353 on Build GRCh38
rs527957641 Chr.15: 40767402-40767402 on Build GRCh38
rs970281700 Chr.15: 40767348-40767348 on Build GRCh38
rs985303127 Chr.15: 40767465-40767465 on Build GRCh38
rs749155056 Chr.15: 40767307-40767307 on Build GRCh38
rs531608687 Chr.15: 40767393-40767393 on Build GRCh38
rs953895813 Chr.15: 40767459-40767459 on Build GRCh38
rs765589928 Chr.15: 40767372-40767372 on Build GRCh38
rs562588825 Chr.15: 40767376-40767376 on Build GRCh38
rs1052000044 Chr.15: 40767447-40767447 on Build GRCh38
rs571040859 Chr.15: 40767477-40767477 on Build GRCh38
rs776744478 Chr.15: 40767400-40767400 on Build GRCh38
rs774287216 Chr.15: 40767315-40767315 on Build GRCh38
rs149691221 Chr.15: 40767400-40767400 on Build GRCh38
rs745754620 Chr.15: 40767413-40767413 on Build GRCh38
rs775737051 Chr.15: 40767328-40767328 on Build GRCh38
rs750728810 Chr.15: 40767373-40767373 on Build GRCh38

View Full Product Details