Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 18705730-18706003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF22
Entrez Gene ID
283284
Gene Name
immunoglobulin superfamily member 22
Gene Aliases
IGFN2
UniGene
Hs.434152
Gene Chromosome Location
Chr.11: 18704305-18726939 on Build GRCh38
Transcripts
NM_173588.3 XM_011520030.2 XM_011520031.2 XM_017017607.1 XM_017017608.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3752276 c.3826A>G p.T1276A Chr.11: 18705901-18705901 on Build GRCh38
4647293 c.3750C>T p.D1250D Chr.11: 18705977-18705977 on Build GRCh38
3738984 c.3893G>A p.C1298Y Chr.11: 18705834-18705834 on Build GRCh38
5121967 c.3910+8C>T p.? Chr.11: 18705809-18705809 on Build GRCh38
3979481 c.3796G>T p.G1266C Chr.11: 18705931-18705931 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1020271052 Chr.11: 18705832-18705832 on Build GRCh38
rs770289862 Chr.11: 18705936-18705936 on Build GRCh38
rs1043074536 Chr.11: 18705794-18705794 on Build GRCh38
rs1042241079 Chr.11: 18705818-18705818 on Build GRCh38
rs750037408 Chr.11: 18705844-18705844 on Build GRCh38
rs970785491 Chr.11: 18705940-18705940 on Build GRCh38
rs529627690 Chr.11: 18705943-18705943 on Build GRCh38
rs997439719 Chr.11: 18705840-18705840 on Build GRCh38
rs1011585015 Chr.11: 18705784-18705784 on Build GRCh38
rs562339587 Chr.11: 18705834-18705834 on Build GRCh38
rs753590937 Chr.11: 18705796-18705796 on Build GRCh38
rs999139894 Chr.11: 18705806-18705806 on Build GRCh38
rs781769615 Chr.11: 18705926-18705926 on Build GRCh38
rs890863825 Chr.11: 18705779-18705779 on Build GRCh38
rs149702932 Chr.11: 18705916-18705916 on Build GRCh38
rs766953015 Chr.11: 18705929-18705929 on Build GRCh38
rs538222440 Chr.11: 18705780-18705780 on Build GRCh38
rs779408135 Chr.11: 18705871-18705871 on Build GRCh38
rs903180989 Chr.11: 18705804-18705804 on Build GRCh38
rs868816925 Chr.11: 18705948-18705948 on Build GRCh38
rs546661546 Chr.11: 18705768-18705768 on Build GRCh38
rs571040017 Chr.11: 18705775-18705775 on Build GRCh38
rs533509427 Chr.11: 18705898-18705899 on Build GRCh38
rs1015975738 Chr.11: 18705928-18705928 on Build GRCh38
rs530222528 Chr.11: 18705965-18705965 on Build GRCh38
rs749461925 Chr.11: 18705962-18705962 on Build GRCh38
rs758125393 Chr.11: 18705860-18705860 on Build GRCh38
rs374121144 Chr.11: 18705953-18705953 on Build GRCh38
rs984529900 Chr.11: 18705921-18705921 on Build GRCh38
rs376134827 Chr.11: 18705785-18705785 on Build GRCh38
rs1867877 Chr.11: 18705901-18705901 on Build GRCh38
rs1001155316 Chr.11: 18705859-18705859 on Build GRCh38
rs773799121 Chr.11: 18705819-18705819 on Build GRCh38

View Full Product Details