Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 69383429-69383702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM189A2
Entrez Gene ID
9413
Gene Name
family with sequence similarity 189 member A2
Gene Aliases
C9orf61, X123
UniGene
Hs.118003
Gene Chromosome Location
Chr.9: 69324572-69392461 on Build GRCh38
Transcripts
NM_001127608.1 NM_004816.3 XM_005252307.4 XM_017015324.1 XM_017015325.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1109715 c.535G>A p.G179R Chr.9: 69383670-69383670 on Build GRCh38
5530870 c.542G>A p.R181Q Chr.9: 69383677-69383677 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs137983537 Chr.9: 69383667-69383667 on Build GRCh38
rs749184997 Chr.9: 69383638-69383638 on Build GRCh38
rs375507119 Chr.9: 69383599-69383599 on Build GRCh38
rs780818783 Chr.9: 69383616-69383616 on Build GRCh38
rs1021982809 Chr.9: 69383588-69383588 on Build GRCh38
rs778390225 Chr.9: 69383586-69383586 on Build GRCh38
rs939680584 Chr.9: 69383464-69383464 on Build GRCh38
rs889546581 Chr.9: 69383576-69383576 on Build GRCh38
rs370536130 Chr.9: 69383581-69383581 on Build GRCh38
rs757732064 Chr.9: 69383676-69383676 on Build GRCh38
rs369558547 Chr.9: 69383677-69383677 on Build GRCh38
rs948465296 Chr.9: 69383492-69383492 on Build GRCh38
rs542512596 Chr.9: 69383461-69383461 on Build GRCh38
rs760813310 Chr.9: 69383669-69383669 on Build GRCh38
rs767533038 Chr.9: 69383580-69383580 on Build GRCh38
rs770701747 Chr.9: 69383650-69383650 on Build GRCh38
rs41277919 Chr.9: 69383618-69383618 on Build GRCh38
rs775318543 Chr.9: 69383474-69383474 on Build GRCh38
rs147770155 Chr.9: 69383595-69383595 on Build GRCh38
rs1053994487 Chr.9: 69383584-69383584 on Build GRCh38
rs779162612 Chr.9: 69383602-69383602 on Build GRCh38
rs368747900 Chr.9: 69383610-69383610 on Build GRCh38
rs965753314 Chr.9: 69383625-69383625 on Build GRCh38
rs535780765 Chr.9: 69383462-69383462 on Build GRCh38
rs371522403 Chr.9: 69383598-69383598 on Build GRCh38
rs768454234 Chr.9: 69383611-69383611 on Build GRCh38
rs375093851 Chr.9: 69383619-69383619 on Build GRCh38
rs759709618 Chr.9: 69383662-69383662 on Build GRCh38
rs10780333 Chr.9: 69383475-69383475 on Build GRCh38
rs1056795148 Chr.9: 69383465-69383465 on Build GRCh38
rs755601946 Chr.9: 69383565-69383565 on Build GRCh38
rs774935336 Chr.9: 69383630-69383632 on Build GRCh38
rs149502376 Chr.9: 69383670-69383670 on Build GRCh38
rs777514947 Chr.9: 69383505-69383505 on Build GRCh38
rs118054476 Chr.9: 69383653-69383653 on Build GRCh38
rs112688055 Chr.9: 69383573-69383573 on Build GRCh38
rs756579090 Chr.9: 69383582-69383582 on Build GRCh38
rs1044529488 Chr.9: 69383560-69383560 on Build GRCh38
rs767764158 Chr.9: 69383663-69383663 on Build GRCh38

View Full Product Details