Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 13700150-13700411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAR1
Entrez Gene ID
84188
Gene Name
fatty acyl-CoA reductase 1
Gene Aliases
MLSTD2, PFCRD, SDR10E1
UniGene
Hs.501991
Gene Chromosome Location
Chr.11: 13668659-13732346 on Build GRCh38
Transcripts
NM_032228.5 XM_011520400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1604346 c.248A>G p.N83S Chr.11: 13700375-13700375 on Build GRCh38
3415705 c.233A>T p.K78I Chr.11: 13700360-13700360 on Build GRCh38
6400183 c.254A>G p.E85G Chr.11: 13700381-13700381 on Build GRCh38
3445578 c.220G>T p.D74Y Chr.11: 13700347-13700347 on Build GRCh38
1737648 c.238A>T p.I80L Chr.11: 13700365-13700365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898669242 Chr.11: 13700207-13700207 on Build GRCh38
rs141912736 Chr.11: 13700343-13700343 on Build GRCh38
rs746235708 Chr.11: 13700275-13700275 on Build GRCh38
rs759397446 Chr.11: 13700360-13700360 on Build GRCh38
rs767316052 Chr.11: 13700367-13700367 on Build GRCh38
rs749892986 Chr.11: 13700256-13700256 on Build GRCh38
rs577936184 Chr.11: 13700195-13700195 on Build GRCh38
rs879257611 Chr.11: 13700260-13700260 on Build GRCh38
rs533310795 Chr.11: 13700238-13700238 on Build GRCh38
rs200062896 Chr.11: 13700287-13700288 on Build GRCh38
rs763681183 Chr.11: 13700379-13700379 on Build GRCh38
rs753461426 Chr.11: 13700385-13700385 on Build GRCh38
rs373841275 Chr.11: 13700272-13700272 on Build GRCh38
rs758770625 Chr.11: 13700283-13700283 on Build GRCh38
rs780575823 Chr.11: 13700236-13700236 on Build GRCh38
rs371508883 Chr.11: 13700234-13700234 on Build GRCh38
rs928641356 Chr.11: 13700220-13700220 on Build GRCh38
rs566033321 Chr.11: 13700224-13700224 on Build GRCh38
rs747024417 Chr.11: 13700300-13700300 on Build GRCh38
rs775981545 Chr.11: 13700292-13700294 on Build GRCh38
rs774067231 Chr.11: 13700356-13700356 on Build GRCh38
rs770071264 Chr.11: 13700332-13700332 on Build GRCh38
rs780637729 Chr.11: 13700299-13700299 on Build GRCh38
rs771928820 Chr.11: 13700218-13700219 on Build GRCh38
rs768753013 Chr.11: 13700305-13700305 on Build GRCh38
rs377505176 Chr.11: 13700312-13700312 on Build GRCh38
rs551576122 Chr.11: 13700239-13700239 on Build GRCh38
rs775466274 Chr.11: 13700371-13700371 on Build GRCh38
rs760577650 Chr.11: 13700375-13700375 on Build GRCh38
rs570428526 Chr.11: 13700253-13700253 on Build GRCh38
rs148102733 Chr.11: 13700317-13700317 on Build GRCh38

View Full Product Details