Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75441258-75441499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD5
Entrez Gene ID
81544
Gene Name
glycerophosphodiester phosphodiesterase domain containing 5
Gene Aliases
GDE2, PP1665
UniGene
Hs.503297
Gene Chromosome Location
Chr.11: 75434640-75525916 on Build GRCh38
Transcripts
NM_030792.6 XM_006718697.3 XM_011545276.2 XM_011545277.2 XM_011545278.2 XM_011545279.2 XM_011545280.1 XM_011545281.2 XM_011545285.2 XM_011545286.1 XM_011545287.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6670401 c.1355A>G p.N452S Chr.11: 75441281-75441281 on Build GRCh38
1728207 c.1351G>A p.V451M Chr.11: 75441285-75441285 on Build GRCh38
5562164 c.1376C>T p.P459L Chr.11: 75441260-75441260 on Build GRCh38
5999841 c.1339T>C p.W447R Chr.11: 75441297-75441297 on Build GRCh38
2018027 c.1335G>A p.A445A Chr.11: 75441301-75441301 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749265714 Chr.11: 75441301-75441301 on Build GRCh38
rs139147150 Chr.11: 75441451-75441451 on Build GRCh38
rs775937690 Chr.11: 75441333-75441333 on Build GRCh38
rs746187080 Chr.11: 75441329-75441329 on Build GRCh38
rs376933894 Chr.11: 75441343-75441343 on Build GRCh38
rs1032718394 Chr.11: 75441286-75441286 on Build GRCh38
rs552513006 Chr.11: 75441472-75441472 on Build GRCh38
rs995156801 Chr.11: 75441379-75441379 on Build GRCh38
rs375341676 Chr.11: 75441350-75441350 on Build GRCh38
rs976262214 Chr.11: 75441429-75441429 on Build GRCh38
rs778547647 Chr.11: 75441303-75441303 on Build GRCh38
rs750796873 Chr.11: 75441345-75441345 on Build GRCh38
rs1003383521 Chr.11: 75441367-75441367 on Build GRCh38
rs931654141 Chr.11: 75441428-75441428 on Build GRCh38
rs890355355 Chr.11: 75441432-75441432 on Build GRCh38
rs574141554 Chr.11: 75441433-75441433 on Build GRCh38
rs879687222 Chr.11: 75441373-75441376 on Build GRCh38
rs772801861 Chr.11: 75441322-75441322 on Build GRCh38
rs771630581 Chr.11: 75441452-75441452 on Build GRCh38
rs898177010 Chr.11: 75441334-75441334 on Build GRCh38
rs1000476169 Chr.11: 75441453-75441453 on Build GRCh38
rs150724500 Chr.11: 75441408-75441408 on Build GRCh38
rs113035931 Chr.11: 75441473-75441473 on Build GRCh38
rs982334202 Chr.11: 75441475-75441475 on Build GRCh38
rs560825092 Chr.11: 75441415-75441415 on Build GRCh38
rs1008830627 Chr.11: 75441435-75441435 on Build GRCh38
rs908060176 Chr.11: 75441395-75441395 on Build GRCh38
rs923566773 Chr.11: 75441285-75441285 on Build GRCh38
rs763475051 Chr.11: 75441340-75441340 on Build GRCh38
rs188755757 Chr.11: 75441304-75441304 on Build GRCh38
rs559824925 Chr.11: 75441455-75441455 on Build GRCh38
rs749736058 Chr.11: 75441440-75441440 on Build GRCh38
rs755870440 Chr.11: 75441293-75441293 on Build GRCh38
rs142042920 Chr.11: 75441397-75441397 on Build GRCh38
rs977617658 Chr.11: 75441416-75441416 on Build GRCh38
rs553047306 Chr.11: 75441370-75441370 on Build GRCh38
rs780066269 Chr.11: 75441298-75441298 on Build GRCh38
rs773984780 Chr.11: 75441463-75441463 on Build GRCh38
rs907179647 Chr.11: 75441352-75441352 on Build GRCh38
rs750219895 Chr.11: 75441277-75441277 on Build GRCh38
rs959167634 Chr.11: 75441371-75441371 on Build GRCh38
rs532185515 Chr.11: 75441462-75441462 on Build GRCh38
rs56155408 Chr.11: 75441330-75441330 on Build GRCh38
rs920375229 Chr.11: 75441411-75441411 on Build GRCh38
rs754486109 Chr.11: 75441302-75441302 on Build GRCh38
rs767720380 Chr.11: 75441348-75441348 on Build GRCh38
rs140725155 Chr.11: 75441305-75441305 on Build GRCh38
rs764623624 Chr.11: 75441342-75441342 on Build GRCh38
rs1030636531 Chr.11: 75441419-75441419 on Build GRCh38
rs965728904 Chr.11: 75441424-75441424 on Build GRCh38

View Full Product Details