Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176093473-176093698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXD13
Entrez Gene ID
3239
Gene Name
homeobox D13
Gene Aliases
BDE, BDSD, HOX4I, SPD, SPD1
UniGene
Hs.152414
Gene Chromosome Location
Chr.2: 176087487-176095938 on Build GRCh38
Transcripts
NM_000523.3 XM_011511068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7098991 c.609G>T p.V203V Chr.2: 176093499-176093499 on Build GRCh38
6510037 c.726G>A p.Q242Q Chr.2: 176093641-176093641 on Build GRCh38
6857092 c.729C>T p.Y243Y Chr.2: 176093619-176093619 on Build GRCh38
1305872 c.624G>A p.G208G Chr.2: 176093539-176093539 on Build GRCh38
5567790 c.718A>C p.S240R Chr.2: 176093608-176093608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs878854400 Chr.2: 176093632-176093632 on Build GRCh38
rs749474029 Chr.2: 176093515-176093515 on Build GRCh38
rs776131391 Chr.2: 176093601-176093601 on Build GRCh38
rs779934918 Chr.2: 176093495-176093495 on Build GRCh38
rs374920154 Chr.2: 176093647-176093647 on Build GRCh38
rs200274374 Chr.2: 176093519-176093519 on Build GRCh38
rs566987103 Chr.2: 176093505-176093505 on Build GRCh38
rs764371902 Chr.2: 176093613-176093613 on Build GRCh38
rs577016423 Chr.2: 176093576-176093576 on Build GRCh38
rs917075646 Chr.2: 176093639-176093639 on Build GRCh38
rs544065459 Chr.2: 176093628-176093628 on Build GRCh38
rs746854273 Chr.2: 176093503-176093503 on Build GRCh38
rs367674921 Chr.2: 176093564-176093564 on Build GRCh38
rs121912541 Chr.2: 176093573-176093573 on Build GRCh38
rs575180284 Chr.2: 176093615-176093615 on Build GRCh38
rs774314169 Chr.2: 176093560-176093560 on Build GRCh38
rs778122220 Chr.2: 176093509-176093509 on Build GRCh38
rs771123279 Chr.2: 176093532-176093532 on Build GRCh38
rs761394795 Chr.2: 176093624-176093624 on Build GRCh38
rs147720746 Chr.2: 176093507-176093507 on Build GRCh38
rs35290213 Chr.2: 176093644-176093644 on Build GRCh38
rs776997927 Chr.2: 176093622-176093622 on Build GRCh38
rs1009388093 Chr.2: 176093675-176093675 on Build GRCh38
rs749975299 Chr.2: 176093630-176093630 on Build GRCh38
rs886037831 Chr.2: 176093672-176093672 on Build GRCh38
rs1016989207 Chr.2: 176093528-176093528 on Build GRCh38
rs142600246 Chr.2: 176093580-176093580 on Build GRCh38
rs1047620716 Chr.2: 176093500-176093500 on Build GRCh38
rs751492206 Chr.2: 176093679-176093679 on Build GRCh38
rs751321734 Chr.2: 176093631-176093631 on Build GRCh38
rs556455717 Chr.2: 176093645-176093645 on Build GRCh38
rs761192563 Chr.2: 176093612-176093612 on Build GRCh38

View Full Product Details