Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127507387-127507642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM129B
Entrez Gene ID
64855
Gene Name
family with sequence similarity 129 member B
Gene Aliases
C9orf88, MEG-3, MINERVA, OC58, bA356B19.6
UniGene
Hs.522401
Gene Chromosome Location
Chr.9: 127505338-127579007 on Build GRCh38
Transcripts
NM_001035534.2 NM_022833.3 XM_005252135.2 XM_011518925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4682515 c.1695C>T p.Y565Y Chr.9: 127507391-127507391 on Build GRCh38
6568757 c.1624G>A p.E542K Chr.9: 127507423-127507423 on Build GRCh38
4682516 c.1656C>T p.Y552Y Chr.9: 127507391-127507391 on Build GRCh38
6568756 c.1663G>A p.E555K Chr.9: 127507423-127507423 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs973569318 Chr.9: 127507556-127507556 on Build GRCh38
rs1010714502 Chr.9: 127507423-127507423 on Build GRCh38
rs772785297 Chr.9: 127507453-127507453 on Build GRCh38
rs111318680 Chr.9: 127507433-127507433 on Build GRCh38
rs2245051 Chr.9: 127507507-127507507 on Build GRCh38
rs557020664 Chr.9: 127507567-127507567 on Build GRCh38
rs947745981 Chr.9: 127507608-127507608 on Build GRCh38
rs745937259 Chr.9: 127507590-127507590 on Build GRCh38
rs147775231 Chr.9: 127507427-127507427 on Build GRCh38
rs771249314 Chr.9: 127507417-127507417 on Build GRCh38
rs548488288 Chr.9: 127507514-127507514 on Build GRCh38
rs534944272 Chr.9: 127507513-127507513 on Build GRCh38
rs999167607 Chr.9: 127507512-127507512 on Build GRCh38
rs568275695 Chr.9: 127507533-127507533 on Build GRCh38
rs561975739 Chr.9: 127507410-127507410 on Build GRCh38
rs564397558 Chr.9: 127507448-127507448 on Build GRCh38
rs530272537 Chr.9: 127507418-127507418 on Build GRCh38
rs747725150 Chr.9: 127507445-127507445 on Build GRCh38
rs759413188 Chr.9: 127507480-127507480 on Build GRCh38
rs570413120 Chr.9: 127507587-127507587 on Build GRCh38
rs776493795 Chr.9: 127507467-127507467 on Build GRCh38
rs374268088 Chr.9: 127507437-127507437 on Build GRCh38
rs999847409 Chr.9: 127507414-127507414 on Build GRCh38
rs908061374 Chr.9: 127507523-127507523 on Build GRCh38
rs746943461 Chr.9: 127507498-127507498 on Build GRCh38
rs767092510 Chr.9: 127507481-127507481 on Build GRCh38
rs369527501 Chr.9: 127507415-127507415 on Build GRCh38
rs991096088 Chr.9: 127507571-127507571 on Build GRCh38
rs192289530 Chr.9: 127507551-127507551 on Build GRCh38
rs539512361 Chr.9: 127507588-127507588 on Build GRCh38
rs188468532 Chr.9: 127507499-127507499 on Build GRCh38
rs759451839 Chr.9: 127507495-127507496 on Build GRCh38
rs3739814 Chr.9: 127507465-127507465 on Build GRCh38
rs773982771 Chr.9: 127507439-127507439 on Build GRCh38
rs760394527 Chr.9: 127507461-127507461 on Build GRCh38
rs964730229 Chr.9: 127507431-127507431 on Build GRCh38
rs893611586 Chr.9: 127507422-127507422 on Build GRCh38
rs530994276 Chr.9: 127507416-127507416 on Build GRCh38
rs771816596 Chr.9: 127507451-127507451 on Build GRCh38

View Full Product Details