Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41821615-41821888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP10
Entrez Gene ID
60681
Gene Name
FK506 binding protein 10
Gene Aliases
BRKS1, FKBP65, OI11, OI6, PPIASE, hFKBP65
UniGene
Hs.463035
Gene Chromosome Location
Chr.17: 41812262-41823217 on Build GRCh38
Transcripts
NM_021939.3 XM_011525099.2 XM_011525100.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6146472 c.1540G>T p.D514Y Chr.17: 41821794-41821794 on Build GRCh38
4066451 c.748C>T p.L250L Chr.17: 41821683-41821683 on Build GRCh38
5386368 c.820C>T p.P274S Chr.17: 41821755-41821755 on Build GRCh38
6146470 c.1545C>A p.G515G Chr.17: 41821799-41821799 on Build GRCh38
353948 c.1557G>A p.P519P Chr.17: 41821811-41821811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782187683 Chr.17: 41821806-41821806 on Build GRCh38
rs782168569 Chr.17: 41821833-41821833 on Build GRCh38
rs782378575 Chr.17: 41821644-41821644 on Build GRCh38
rs143556345 Chr.17: 41821733-41821733 on Build GRCh38
rs782574252 Chr.17: 41821866-41821866 on Build GRCh38
rs782532142 Chr.17: 41821730-41821731 on Build GRCh38
rs144314381 Chr.17: 41821708-41821708 on Build GRCh38
rs112668233 Chr.17: 41821803-41821803 on Build GRCh38
rs542334549 Chr.17: 41821656-41821656 on Build GRCh38
rs541713038 Chr.17: 41821811-41821811 on Build GRCh38
rs782276179 Chr.17: 41821647-41821647 on Build GRCh38
rs117224218 Chr.17: 41821843-41821843 on Build GRCh38
rs781822714 Chr.17: 41821730-41821730 on Build GRCh38
rs199997285 Chr.17: 41821777-41821777 on Build GRCh38
rs541344214 Chr.17: 41821799-41821799 on Build GRCh38
rs782528448 Chr.17: 41821859-41821859 on Build GRCh38
rs781930058 Chr.17: 41821707-41821707 on Build GRCh38
rs371667550 Chr.17: 41821861-41821861 on Build GRCh38
rs782755952 Chr.17: 41821735-41821735 on Build GRCh38
rs782082258 Chr.17: 41821829-41821829 on Build GRCh38
rs182545136 Chr.17: 41821739-41821739 on Build GRCh38
rs782743762 Chr.17: 41821853-41821853 on Build GRCh38
rs782259238 Chr.17: 41821641-41821641 on Build GRCh38
rs782331462 Chr.17: 41821648-41821648 on Build GRCh38
rs782531690 Chr.17: 41821770-41821770 on Build GRCh38
rs782742068 Chr.17: 41821729-41821729 on Build GRCh38
rs923789200 Chr.17: 41821637-41821637 on Build GRCh38
rs894767306 Chr.17: 41821822-41821822 on Build GRCh38
rs782442135 Chr.17: 41821862-41821862 on Build GRCh38
rs903491212 Chr.17: 41821854-41821854 on Build GRCh38
rs782810622 Chr.17: 41821834-41821834 on Build GRCh38
rs797044794 Chr.17: 41821804-41821804 on Build GRCh38
rs781987497 Chr.17: 41821832-41821832 on Build GRCh38
rs782611610 Chr.17: 41821792-41821792 on Build GRCh38
rs764950164 Chr.17: 41821679-41821679 on Build GRCh38
rs368439289 Chr.17: 41821860-41821860 on Build GRCh38
rs781978984 Chr.17: 41821665-41821665 on Build GRCh38
rs782455799 Chr.17: 41821846-41821846 on Build GRCh38
rs782180826 Chr.17: 41821780-41821780 on Build GRCh38
rs140883152 Chr.17: 41821810-41821810 on Build GRCh38
rs782603490 Chr.17: 41821669-41821669 on Build GRCh38
rs114199361 Chr.17: 41821650-41821650 on Build GRCh38
rs782776800 Chr.17: 41821831-41821831 on Build GRCh38
rs76022961 Chr.17: 41821800-41821800 on Build GRCh38
rs782108520 Chr.17: 41821655-41821655 on Build GRCh38
rs782152511 Chr.17: 41821668-41821668 on Build GRCh38

View Full Product Details