Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49136267-49136494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC3A
Entrez Gene ID
22862
Gene Name
fibronectin type III domain containing 3A
Gene Aliases
FNDC3, HUGO, bA203I16.1, bA203I16.5
UniGene
Hs.508010
Gene Chromosome Location
Chr.13: 48975241-49209779 on Build GRCh38
Transcripts
NM_001079673.1 NM_001278438.1 NM_014923.4 NR_103528.1 XM_011534997.2 XM_017020440.1 XM_017020441.1 XM_017020442.1 XM_017020443.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
947828 c.533G>A p.R178Q Chr.13: 49136374-49136374 on Build GRCh38
1367352 c.578G>A p.R193H Chr.13: 49136419-49136419 on Build GRCh38
1730687 c.650A>C p.E217A Chr.13: 49136491-49136491 on Build GRCh38
6665880 c.561_562insA p.L190fs*11 Chr.13: 49136402-49136403 on Build GRCh38
1677662 c.532C>T p.R178* Chr.13: 49136373-49136373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766771306 Chr.13: 49136364-49136364 on Build GRCh38
rs201895066 Chr.13: 49136313-49136313 on Build GRCh38
rs542056437 Chr.13: 49136322-49136322 on Build GRCh38
rs555788664 Chr.13: 49136410-49136410 on Build GRCh38
rs756811491 Chr.13: 49136324-49136324 on Build GRCh38
rs374005060 Chr.13: 49136447-49136447 on Build GRCh38
rs759935299 Chr.13: 49136379-49136379 on Build GRCh38
rs747008152 Chr.13: 49136444-49136444 on Build GRCh38
rs773384682 Chr.13: 49136443-49136443 on Build GRCh38
rs751001700 Chr.13: 49136390-49136390 on Build GRCh38
rs745693809 Chr.13: 49136408-49136408 on Build GRCh38
rs755840596 Chr.13: 49136409-49136409 on Build GRCh38
rs780944057 Chr.13: 49136404-49136404 on Build GRCh38
rs752339500 Chr.13: 49136387-49136387 on Build GRCh38
rs1003943137 Chr.13: 49136386-49136386 on Build GRCh38
rs777155005 Chr.13: 49136374-49136374 on Build GRCh38
rs373831224 Chr.13: 49136337-49136337 on Build GRCh38
rs573953502 Chr.13: 49136446-49136446 on Build GRCh38
rs745417628 Chr.13: 49136359-49136359 on Build GRCh38
rs1033619369 Chr.13: 49136457-49136457 on Build GRCh38
rs770767033 Chr.13: 49136308-49136308 on Build GRCh38
rs772136953 Chr.13: 49136340-49136340 on Build GRCh38
rs773154802 Chr.13: 49136356-49136356 on Build GRCh38
rs768889396 Chr.13: 49136335-49136335 on Build GRCh38
rs994532565 Chr.13: 49136435-49136435 on Build GRCh38
rs757270122 Chr.13: 49136395-49136395 on Build GRCh38
rs376763239 Chr.13: 49136389-49136389 on Build GRCh38
rs747023839 Chr.13: 49136297-49136297 on Build GRCh38
rs761890157 Chr.13: 49136339-49136339 on Build GRCh38
rs745642614 Chr.13: 49136327-49136327 on Build GRCh38
rs759992285 Chr.13: 49136452-49136452 on Build GRCh38
rs764982022 Chr.13: 49136381-49136381 on Build GRCh38
rs369583924 Chr.13: 49136419-49136419 on Build GRCh38
rs111902916 Chr.13: 49136456-49136456 on Build GRCh38
rs778364556 Chr.13: 49136412-49136412 on Build GRCh38

View Full Product Details