Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 117543609-117543868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMX2
Entrez Gene ID
2018
Gene Name
empty spiracles homeobox 2
Gene Aliases
-
UniGene
Hs.202095
Gene Chromosome Location
Chr.10: 117542445-117549546 on Build GRCh38
Transcripts
NM_001165924.1 NM_004098.3 NR_002791.2

Gene Symbol
EMX2OS
Entrez Gene ID
196047
Gene Name
EMX2 opposite strand/antisense RNA
Gene Aliases
EMX2-AS1, NCRNA00045
UniGene
Hs.312592
Gene Chromosome Location
Chr.10: 117484293-117545068 on Build GRCh38
Transcripts
NM_001165924.1 NM_004098.3 NR_002791.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
239701 c.352A>C p.T118P Chr.10: 117543619-117543619 on Build GRCh38
7173625 c.394C>T p.H132Y Chr.10: 117543661-117543661 on Build GRCh38
6049180 c.348G>T p.P116P Chr.10: 117543615-117543615 on Build GRCh38
5567279 c.351C>T p.S117S Chr.10: 117543618-117543618 on Build GRCh38
7226865 c.358T>C p.Y120H Chr.10: 117543625-117543625 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537483924 Chr.10: 117543777-117543777 on Build GRCh38
rs749867034 Chr.10: 117543712-117543712 on Build GRCh38
rs148439570 Chr.10: 117543642-117543642 on Build GRCh38
rs1047735740 Chr.10: 117543832-117543832 on Build GRCh38
rs565883191 Chr.10: 117543668-117543668 on Build GRCh38
rs766457972 Chr.10: 117543703-117543703 on Build GRCh38
rs995647407 Chr.10: 117543747-117543747 on Build GRCh38
rs375505457 Chr.10: 117543695-117543695 on Build GRCh38
rs770214262 Chr.10: 117543743-117543743 on Build GRCh38
rs751473437 Chr.10: 117543708-117543708 on Build GRCh38
rs779534951 Chr.10: 117543722-117543722 on Build GRCh38
rs965415571 Chr.10: 117543806-117543806 on Build GRCh38
rs975865088 Chr.10: 117543835-117543835 on Build GRCh38
rs767010028 Chr.10: 117543711-117543711 on Build GRCh38
rs555087239 Chr.10: 117543705-117543705 on Build GRCh38
rs756817634 Chr.10: 117543710-117543710 on Build GRCh38
rs370645871 Chr.10: 117543700-117543700 on Build GRCh38
rs377401555 Chr.10: 117543729-117543729 on Build GRCh38
rs568350366 Chr.10: 117543707-117543707 on Build GRCh38
rs796837297 Chr.10: 117543836-117543836 on Build GRCh38
rs780867496 Chr.10: 117543644-117543644 on Build GRCh38
rs1054888364 Chr.10: 117543742-117543742 on Build GRCh38
rs749703700 Chr.10: 117543724-117543724 on Build GRCh38
rs745507404 Chr.10: 117543652-117543652 on Build GRCh38
rs888724443 Chr.10: 117543833-117543833 on Build GRCh38
rs371316386 Chr.10: 117543687-117543687 on Build GRCh38
rs1004214830 Chr.10: 117543788-117543788 on Build GRCh38
rs770122962 Chr.10: 117543661-117543661 on Build GRCh38
rs936472300 Chr.10: 117543764-117543764 on Build GRCh38
rs1002394195 Chr.10: 117543726-117543726 on Build GRCh38
rs777519993 Chr.10: 117543628-117543628 on Build GRCh38
rs751097554 Chr.10: 117543640-117543640 on Build GRCh38
rs8192645 Chr.10: 117543840-117543840 on Build GRCh38
rs769239455 Chr.10: 117543688-117543688 on Build GRCh38
rs374844113 Chr.10: 117543778-117543778 on Build GRCh38
rs774967972 Chr.10: 117543691-117543691 on Build GRCh38
rs113636926 Chr.10: 117543798-117543798 on Build GRCh38
rs755802506 Chr.10: 117543715-117543715 on Build GRCh38
rs8192644 Chr.10: 117543786-117543786 on Build GRCh38
rs929654999 Chr.10: 117543827-117543827 on Build GRCh38
rs368291354 Chr.10: 117543697-117543697 on Build GRCh38

View Full Product Details