Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 206704792-206705029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYTN
Entrez Gene ID
391475
Gene Name
dystrotelin
Gene Aliases
-
UniGene
Hs.640667
Gene Chromosome Location
Chr.2: 206651621-206718396 on Build GRCh38
Transcripts
NM_001093730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6876646 c.441G>A p.M147I Chr.2: 206704885-206704885 on Build GRCh38
337098 c.466C>G p.L156V Chr.2: 206704860-206704860 on Build GRCh38
4969599 c.411C>A p.S137R Chr.2: 206704915-206704915 on Build GRCh38
1015468 c.386T>C p.L129P Chr.2: 206704940-206704940 on Build GRCh38
5530287 c.458G>A p.R153K Chr.2: 206704868-206704868 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532282575 Chr.2: 206704953-206704953 on Build GRCh38
rs764117520 Chr.2: 206704837-206704837 on Build GRCh38
rs780940766 Chr.2: 206704849-206704849 on Build GRCh38
rs746142719 Chr.2: 206704854-206704854 on Build GRCh38
rs114675847 Chr.2: 206704856-206704856 on Build GRCh38
rs371211980 Chr.2: 206704861-206704861 on Build GRCh38
rs200720001 Chr.2: 206704916-206704916 on Build GRCh38
rs749429198 Chr.2: 206704912-206704912 on Build GRCh38
rs1046665765 Chr.2: 206704864-206704864 on Build GRCh38
rs750236316 Chr.2: 206704896-206704896 on Build GRCh38
rs760885257 Chr.2: 206704882-206704882 on Build GRCh38
rs751461326 Chr.2: 206704844-206704844 on Build GRCh38
rs201301748 Chr.2: 206704875-206704875 on Build GRCh38
rs16838626 Chr.2: 206704991-206704991 on Build GRCh38
rs376587365 Chr.2: 206704889-206704889 on Build GRCh38
rs924274267 Chr.2: 206704961-206704961 on Build GRCh38
rs186499304 Chr.2: 206704990-206704990 on Build GRCh38
rs560128332 Chr.2: 206704951-206704951 on Build GRCh38
rs764960410 Chr.2: 206704884-206704884 on Build GRCh38
rs748645481 Chr.2: 206704879-206704879 on Build GRCh38
rs576749690 Chr.2: 206704890-206704890 on Build GRCh38
rs552489709 Chr.2: 206704987-206704987 on Build GRCh38
rs1030282712 Chr.2: 206704964-206704964 on Build GRCh38
rs539863272 Chr.2: 206704933-206704933 on Build GRCh38
rs775028898 Chr.2: 206704876-206704876 on Build GRCh38
rs146519194 Chr.2: 206704888-206704888 on Build GRCh38
rs748731602 Chr.2: 206704918-206704918 on Build GRCh38
rs762488204 Chr.2: 206704983-206704983 on Build GRCh38
rs541491610 Chr.2: 206704848-206704848 on Build GRCh38
rs560966818 Chr.2: 206704911-206704911 on Build GRCh38
rs116768218 Chr.2: 206704899-206704899 on Build GRCh38
rs368477015 Chr.2: 206704880-206704880 on Build GRCh38
rs372283021 Chr.2: 206704881-206704881 on Build GRCh38
rs757081159 Chr.2: 206704846-206704846 on Build GRCh38
rs775251134 Chr.2: 206704981-206704981 on Build GRCh38
rs774959200 Chr.2: 206704885-206704885 on Build GRCh38
rs762760396 Chr.2: 206704973-206704973 on Build GRCh38
rs959739181 Chr.2: 206704973-206704973 on Build GRCh38
rs961928748 Chr.2: 206704944-206704944 on Build GRCh38

View Full Product Details