Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 28058963-28059200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF18A
Entrez Gene ID
81930
Gene Name
kinesin family member 18A
Gene Aliases
MS-KIF18A, PPP1R99
UniGene
Hs.301052
Gene Chromosome Location
Chr.11: 28020616-28108199 on Build GRCh38
Transcripts
NM_031217.3 XM_017018379.1 XM_017018380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
687621 c.1741A>G p.R581G Chr.11: 28059133-28059133 on Build GRCh38
4165754 c.1801T>A p.F601I Chr.11: 28059073-28059073 on Build GRCh38
3935432 c.1881C>G p.N627K Chr.11: 28058993-28058993 on Build GRCh38
5360944 c.1769C>A p.A590D Chr.11: 28059105-28059105 on Build GRCh38
7313525 c.1787C>A p.A596D Chr.11: 28059087-28059087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753811567 Chr.11: 28058998-28058998 on Build GRCh38
rs770059686 Chr.11: 28059069-28059069 on Build GRCh38
rs756594556 Chr.11: 28059150-28059150 on Build GRCh38
rs779654771 Chr.11: 28059029-28059029 on Build GRCh38
rs771420204 Chr.11: 28059164-28059164 on Build GRCh38
rs200977819 Chr.11: 28059062-28059062 on Build GRCh38
rs758718614 Chr.11: 28059016-28059016 on Build GRCh38
rs757992881 Chr.11: 28059023-28059023 on Build GRCh38
rs1023311072 Chr.11: 28059131-28059131 on Build GRCh38
rs767573230 Chr.11: 28059091-28059091 on Build GRCh38
rs183700942 Chr.11: 28059079-28059079 on Build GRCh38
rs776434787 Chr.11: 28059092-28059092 on Build GRCh38
rs370276338 Chr.11: 28059104-28059104 on Build GRCh38
rs765014249 Chr.11: 28059004-28059004 on Build GRCh38
rs139953450 Chr.11: 28059103-28059103 on Build GRCh38
rs918880952 Chr.11: 28059038-28059038 on Build GRCh38
rs779338791 Chr.11: 28059165-28059165 on Build GRCh38
rs764959855 Chr.11: 28059101-28059101 on Build GRCh38
rs780688734 Chr.11: 28059148-28059148 on Build GRCh38
rs773586965 Chr.11: 28059074-28059074 on Build GRCh38
rs746561656 Chr.11: 28059043-28059043 on Build GRCh38
rs774687614 Chr.11: 28059088-28059088 on Build GRCh38
rs754478344 Chr.11: 28059061-28059061 on Build GRCh38
rs73434496 Chr.11: 28059093-28059093 on Build GRCh38
rs896144916 Chr.11: 28059098-28059098 on Build GRCh38
rs146974259 Chr.11: 28059010-28059010 on Build GRCh38
rs778123882 Chr.11: 28059151-28059151 on Build GRCh38
rs985943353 Chr.11: 28058989-28058989 on Build GRCh38
rs572528088 Chr.11: 28059083-28059083 on Build GRCh38
rs754498886 Chr.11: 28059138-28059138 on Build GRCh38
rs764168501 Chr.11: 28058992-28058992 on Build GRCh38
rs749714559 Chr.11: 28059156-28059156 on Build GRCh38
rs751197728 Chr.11: 28059119-28059119 on Build GRCh38
rs757015125 Chr.11: 28058999-28058999 on Build GRCh38

View Full Product Details