Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212858809-212859068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLVCR1
Entrez Gene ID
28982
Gene Name
feline leukemia virus subgroup C cellular receptor 1
Gene Aliases
AXPC1, FLVCR, MFSD7B, PCA, PCARP
UniGene
Hs.7055
Gene Chromosome Location
Chr.1: 212858255-212899363 on Build GRCh38
Transcripts
NM_014053.3 XM_011509446.2 XM_011509447.2 XM_011509448.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7077117 c.515G>A p.G172D Chr.1: 212858967-212858967 on Build GRCh38
3418719 c.555C>T p.C185C Chr.1: 212859007-212859007 on Build GRCh38
5805153 c.388A>T p.I130F Chr.1: 212858840-212858840 on Build GRCh38
4027957 c.366C>T p.A122A Chr.1: 212858818-212858818 on Build GRCh38
3483183 c.372G>A p.Q124Q Chr.1: 212858824-212858824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769501862 Chr.1: 212858917-212858917 on Build GRCh38
rs367576230 Chr.1: 212858852-212858852 on Build GRCh38
rs758893792 Chr.1: 212859003-212859003 on Build GRCh38
rs1053273099 Chr.1: 212858992-212858992 on Build GRCh38
rs772126483 Chr.1: 212858873-212858873 on Build GRCh38
rs764180645 Chr.1: 212858938-212858938 on Build GRCh38
rs779426165 Chr.1: 212858868-212858868 on Build GRCh38
rs753192864 Chr.1: 212858995-212858995 on Build GRCh38
rs750063360 Chr.1: 212858968-212858968 on Build GRCh38
rs747150230 Chr.1: 212858885-212858885 on Build GRCh38
rs886043715 Chr.1: 212858953-212858953 on Build GRCh38
rs751107051 Chr.1: 212858959-212858959 on Build GRCh38
rs369434267 Chr.1: 212858940-212858940 on Build GRCh38
rs925929027 Chr.1: 212858974-212858974 on Build GRCh38
rs767295580 Chr.1: 212858963-212858963 on Build GRCh38
rs779152197 Chr.1: 212858984-212858984 on Build GRCh38
rs751566382 Chr.1: 212859041-212859041 on Build GRCh38
rs994975681 Chr.1: 212859035-212859035 on Build GRCh38
rs903525615 Chr.1: 212859007-212859007 on Build GRCh38
rs757381190 Chr.1: 212859013-212859013 on Build GRCh38
rs886045925 Chr.1: 212858986-212858986 on Build GRCh38
rs374496790 Chr.1: 212859046-212859046 on Build GRCh38
rs755329362 Chr.1: 212858977-212858977 on Build GRCh38
rs371271294 Chr.1: 212858899-212858899 on Build GRCh38
rs762809105 Chr.1: 212858937-212858937 on Build GRCh38
rs142384149 Chr.1: 212859030-212859030 on Build GRCh38
rs144646325 Chr.1: 212858850-212858850 on Build GRCh38
rs781057835 Chr.1: 212858837-212858837 on Build GRCh38
rs145380701 Chr.1: 212859029-212859029 on Build GRCh38
rs201356865 Chr.1: 212858926-212858926 on Build GRCh38
rs140233942 Chr.1: 212859015-212859015 on Build GRCh38
rs892356474 Chr.1: 212858914-212858914 on Build GRCh38
rs41296694 Chr.1: 212859047-212859047 on Build GRCh38
rs200974542 Chr.1: 212858875-212858875 on Build GRCh38
rs775427410 Chr.1: 212858935-212858935 on Build GRCh38
rs759356598 Chr.1: 212858916-212858916 on Build GRCh38
rs761448723 Chr.1: 212858961-212858961 on Build GRCh38
rs776883863 Chr.1: 212858907-212858907 on Build GRCh38
rs267606821 Chr.1: 212859026-212859026 on Build GRCh38
rs756953485 Chr.1: 212858836-212858836 on Build GRCh38
rs747064078 Chr.1: 212859006-212859006 on Build GRCh38

View Full Product Details