Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 50445591-50445844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIGNL1
Entrez Gene ID
63979
Gene Name
fidgetin like 1
Gene Aliases
-
UniGene
Hs.137516
Gene Chromosome Location
Chr.7: 50437327-50450390 on Build GRCh38
Transcripts
NM_001042762.2 NM_001287492.1 NM_001287493.1 NM_001287494.1 NM_001287495.1 NM_001287496.1 NM_022116.4 XM_011515470.2 XM_017012501.1 XM_017012502.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3832978 c.1511C>T p.S504F Chr.7: 50445777-50445777 on Build GRCh38
1313161 c.1567G>A p.E523K Chr.7: 50445721-50445721 on Build GRCh38
6664094 c.1674A>C p.R558S Chr.7: 50445614-50445614 on Build GRCh38
4880734 c.1599A>G p.T533T Chr.7: 50445689-50445689 on Build GRCh38
7140958 c.1639C>A p.R547R Chr.7: 50445649-50445649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927396766 Chr.7: 50445679-50445679 on Build GRCh38
rs751756546 Chr.7: 50445630-50445630 on Build GRCh38
rs546235904 Chr.7: 50445621-50445621 on Build GRCh38
rs753110060 Chr.7: 50445709-50445709 on Build GRCh38
rs751767519 Chr.7: 50445696-50445696 on Build GRCh38
rs779587801 Chr.7: 50445741-50445741 on Build GRCh38
rs778633646 Chr.7: 50445644-50445644 on Build GRCh38
rs202105913 Chr.7: 50445783-50445783 on Build GRCh38
rs767785545 Chr.7: 50445819-50445819 on Build GRCh38
rs1004860572 Chr.7: 50445658-50445658 on Build GRCh38
rs768348091 Chr.7: 50445747-50445747 on Build GRCh38
rs142867266 Chr.7: 50445702-50445702 on Build GRCh38
rs755328374 Chr.7: 50445633-50445633 on Build GRCh38
rs770412510 Chr.7: 50445643-50445643 on Build GRCh38
rs374642968 Chr.7: 50445670-50445670 on Build GRCh38
rs937637560 Chr.7: 50445765-50445765 on Build GRCh38
rs767830198 Chr.7: 50445703-50445703 on Build GRCh38
rs187462087 Chr.7: 50445695-50445695 on Build GRCh38
rs781587853 Chr.7: 50445635-50445635 on Build GRCh38
rs867297337 Chr.7: 50445738-50445738 on Build GRCh38
rs748715277 Chr.7: 50445637-50445637 on Build GRCh38
rs769529142 Chr.7: 50445768-50445768 on Build GRCh38
rs766417250 Chr.7: 50445796-50445796 on Build GRCh38
rs151204868 Chr.7: 50445712-50445712 on Build GRCh38
rs1024229942 Chr.7: 50445665-50445665 on Build GRCh38
rs765160181 Chr.7: 50445681-50445681 on Build GRCh38
rs776196459 Chr.7: 50445759-50445759 on Build GRCh38
rs768379532 Chr.7: 50445675-50445675 on Build GRCh38
rs957806270 Chr.7: 50445689-50445689 on Build GRCh38
rs756481261 Chr.7: 50445711-50445711 on Build GRCh38
rs761804249 Chr.7: 50445678-50445678 on Build GRCh38
rs770578897 Chr.7: 50445632-50445632 on Build GRCh38
rs535171403 Chr.7: 50445765-50445769 on Build GRCh38
rs927756635 Chr.7: 50445763-50445763 on Build GRCh38
rs780152080 Chr.7: 50445622-50445622 on Build GRCh38
rs775032569 Chr.7: 50445652-50445652 on Build GRCh38
rs746692997 Chr.7: 50445744-50445744 on Build GRCh38
rs763041282 Chr.7: 50445692-50445692 on Build GRCh38
rs747945823 Chr.7: 50445764-50445764 on Build GRCh38
rs913508694 Chr.7: 50445775-50445775 on Build GRCh38
rs200644774 Chr.7: 50445810-50445810 on Build GRCh38
rs773072079 Chr.7: 50445687-50445687 on Build GRCh38
rs762844276 Chr.7: 50445788-50445788 on Build GRCh38
rs199544416 Chr.7: 50445676-50445676 on Build GRCh38
rs369897372 Chr.7: 50445714-50445714 on Build GRCh38
rs148546983 Chr.7: 50445649-50445649 on Build GRCh38
rs774478588 Chr.7: 50445798-50445798 on Build GRCh38

View Full Product Details