Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 197050696-197050935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F13B
Entrez Gene ID
2165
Gene Name
coagulation factor XIII B chain
Gene Aliases
FXIIIB
UniGene
Hs.435782
Gene Chromosome Location
Chr.1: 197038741-197067267 on Build GRCh38
Transcripts
NM_001994.2 XM_011509283.2 XM_011509284.2 XM_011509286.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2125172 c.1577C>A p.S526Y Chr.1: 197050858-197050858 on Build GRCh38
3863977 c.1690G>A p.E564K Chr.1: 197050745-197050745 on Build GRCh38
6060759 c.1599A>T p.G533G Chr.1: 197050836-197050836 on Build GRCh38
3863978 c.1682G>A p.G561E Chr.1: 197050753-197050753 on Build GRCh38
6060760 c.1695C>T p.A565A Chr.1: 197050740-197050740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995125530 Chr.1: 197050737-197050737 on Build GRCh38
rs149088047 Chr.1: 197050892-197050892 on Build GRCh38
rs755690618 Chr.1: 197050818-197050818 on Build GRCh38
rs1017292040 Chr.1: 197050786-197050786 on Build GRCh38
rs17549671 Chr.1: 197050849-197050849 on Build GRCh38
rs748731937 Chr.1: 197050745-197050745 on Build GRCh38
rs200450548 Chr.1: 197050791-197050791 on Build GRCh38
rs767545927 Chr.1: 197050809-197050809 on Build GRCh38
rs1026731950 Chr.1: 197050738-197050738 on Build GRCh38
rs367592896 Chr.1: 197050861-197050861 on Build GRCh38
rs145703189 Chr.1: 197050866-197050866 on Build GRCh38
rs759631235 Chr.1: 197050765-197050765 on Build GRCh38
rs6000 Chr.1: 197050728-197050728 on Build GRCh38
rs773338433 Chr.1: 197050750-197050750 on Build GRCh38
rs779125550 Chr.1: 197050735-197050735 on Build GRCh38
rs765945622 Chr.1: 197050832-197050832 on Build GRCh38
rs778260445 Chr.1: 197050862-197050862 on Build GRCh38
rs6001 Chr.1: 197050807-197050807 on Build GRCh38
rs374447970 Chr.1: 197050888-197050888 on Build GRCh38
rs199871990 Chr.1: 197050860-197050860 on Build GRCh38
rs754557020 Chr.1: 197050850-197050850 on Build GRCh38
rs779402912 Chr.1: 197050853-197050853 on Build GRCh38
rs1009314384 Chr.1: 197050800-197050800 on Build GRCh38
rs776673135 Chr.1: 197050867-197050867 on Build GRCh38
rs869261767 Chr.1: 197050893-197050893 on Build GRCh38
rs747109019 Chr.1: 197050753-197050753 on Build GRCh38
rs901737297 Chr.1: 197050835-197050835 on Build GRCh38
rs1032868091 Chr.1: 197050854-197050854 on Build GRCh38
rs772452914 Chr.1: 197050746-197050746 on Build GRCh38
rs750327846 Chr.1: 197050811-197050811 on Build GRCh38
rs113022833 Chr.1: 197050824-197050824 on Build GRCh38
rs547842597 Chr.1: 197050759-197050759 on Build GRCh38
rs571095791 Chr.1: 197050891-197050891 on Build GRCh38
rs747479428 Chr.1: 197050863-197050863 on Build GRCh38

View Full Product Details