Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171281917-171282180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO1
Entrez Gene ID
2326
Gene Name
flavin containing monooxygenase 1
Gene Aliases
-
UniGene
Hs.1424
Gene Chromosome Location
Chr.1: 171248471-171285978 on Build GRCh38
Transcripts
NM_001282692.1 NM_001282693.1 NM_001282694.1 NM_002021.2 XM_005245037.3 XM_005245038.3 XM_006711241.3 XM_006711242.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7308944 c.906C>T p.S302S Chr.1: 171282056-171282056 on Build GRCh38
161110 c.1015G>A p.D339N Chr.1: 171282165-171282165 on Build GRCh38
4913107 c.931G>T p.V311F Chr.1: 171282081-171282081 on Build GRCh38
5957897 c.840A>T p.K280N Chr.1: 171281990-171281990 on Build GRCh38
6665606 c.976G>A p.V326I Chr.1: 171282126-171282126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780421691 Chr.1: 171281956-171281956 on Build GRCh38
rs758875452 Chr.1: 171282089-171282090 on Build GRCh38
rs16864314 Chr.1: 171282057-171282057 on Build GRCh38
rs367883794 Chr.1: 171282107-171282107 on Build GRCh38
rs770436410 Chr.1: 171282083-171282083 on Build GRCh38
rs765757747 Chr.1: 171282077-171282078 on Build GRCh38
rs770468565 Chr.1: 171281997-171281997 on Build GRCh38
rs762916050 Chr.1: 171282026-171282026 on Build GRCh38
rs147725946 Chr.1: 171282071-171282071 on Build GRCh38
rs749740804 Chr.1: 171281958-171281958 on Build GRCh38
rs745465191 Chr.1: 171282094-171282094 on Build GRCh38
rs746192401 Chr.1: 171281942-171281942 on Build GRCh38
rs775420124 Chr.1: 171282133-171282133 on Build GRCh38
rs764096863 Chr.1: 171282155-171282155 on Build GRCh38
rs746472192 Chr.1: 171282077-171282077 on Build GRCh38
rs374416986 Chr.1: 171282036-171282036 on Build GRCh38
rs776028871 Chr.1: 171282092-171282092 on Build GRCh38
rs779522621 Chr.1: 171282076-171282076 on Build GRCh38
rs764194713 Chr.1: 171282030-171282030 on Build GRCh38
rs780333618 Chr.1: 171282063-171282063 on Build GRCh38
rs376072428 Chr.1: 171281967-171281967 on Build GRCh38
rs940240685 Chr.1: 171282093-171282093 on Build GRCh38
rs750892658 Chr.1: 171282080-171282080 on Build GRCh38
rs28360418 Chr.1: 171282058-171282058 on Build GRCh38
rs28360420 Chr.1: 171282129-171282129 on Build GRCh38
rs756344078 Chr.1: 171282059-171282059 on Build GRCh38
rs970563192 Chr.1: 171282032-171282032 on Build GRCh38
rs757403397 Chr.1: 171282055-171282055 on Build GRCh38
rs934223058 Chr.1: 171282040-171282040 on Build GRCh38
rs749722564 Chr.1: 171282068-171282068 on Build GRCh38
rs746587905 Chr.1: 171281981-171281981 on Build GRCh38
rs747318537 Chr.1: 171282093-171282094 on Build GRCh38
rs149030329 Chr.1: 171282139-171282139 on Build GRCh38
rs757271567 Chr.1: 171281949-171281959 on Build GRCh38
rs772709143 Chr.1: 171281976-171281976 on Build GRCh38
rs28360419 Chr.1: 171282114-171282114 on Build GRCh38
rs142868603 Chr.1: 171282022-171282022 on Build GRCh38
rs759144350 Chr.1: 171282017-171282017 on Build GRCh38
rs765073901 Chr.1: 171282021-171282021 on Build GRCh38
rs780521285 Chr.1: 171282092-171282093 on Build GRCh38
rs898942920 Chr.1: 171282127-171282127 on Build GRCh38
rs776387460 Chr.1: 171282010-171282010 on Build GRCh38
rs756539059 Chr.1: 171281945-171281945 on Build GRCh38
rs960580701 Chr.1: 171281995-171281995 on Build GRCh38
rs952203318 Chr.1: 171282123-171282123 on Build GRCh38

View Full Product Details