Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49637512-49637775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMNL3
Entrez Gene ID
91010
Gene Name
formin like 3
Gene Aliases
FHOD3, FRL2, WBP-3, WBP3
UniGene
Hs.179838
Gene Chromosome Location
Chr.12: 49636499-49707414 on Build GRCh38
Transcripts
XM_005269218.2 XM_011538968.2 XM_011538969.2 XM_011538970.2 XM_011538971.2 XM_011538972.2 XM_011538973.2 XM_011538974.2 NM_001031698.2 NM_012272.2 XM_006719324.3 XM_006719325.3 XM_011538135.2 XM_011538136.2 XM_011538137.2 XM_011538138.2 XM_011538139.2 XM_011538140.2 XM_011538141.1 XM_011538143.2 XM_011538144.2 XM_017019135.1 XM_017019136.1 XM_017019137.1 XM_017019138.1 XM_017019139.1

Gene Symbol
PRPF40B
Entrez Gene ID
25766
Gene Name
pre-mRNA processing factor 40 homolog B
Gene Aliases
HYPC
UniGene
Hs.706827
Gene Chromosome Location
Chr.12: 49622717-49644669 on Build GRCh38
Transcripts
XM_005269218.2 XM_011538968.2 XM_011538969.2 XM_011538970.2 XM_011538971.2 XM_011538972.2 XM_011538973.2 XM_011538974.2 NM_001031698.2 NM_012272.2 XM_006719324.3 XM_006719325.3 XM_011538135.2 XM_011538136.2 XM_011538137.2 XM_011538138.2 XM_011538139.2 XM_011538140.2 XM_011538141.1 XM_011538143.2 XM_011538144.2 XM_017019135.1 XM_017019136.1 XM_017019137.1 XM_017019138.1 XM_017019139.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6756611 c.1583G>A p.R528H Chr.12: 49637558-49637558 on Build GRCh38
6434999 c.1610G>A p.G537D Chr.12: 49637733-49637733 on Build GRCh38
4384762 c.1599G>A p.L533L Chr.12: 49637574-49637574 on Build GRCh38
4384761 c.1554G>A p.E518E Chr.12: 49637529-49637529 on Build GRCh38
4806654 c.1566A>C p.A522A Chr.12: 49637541-49637541 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757427271 Chr.12: 49637605-49637605 on Build GRCh38
rs1048430377 Chr.12: 49637709-49637709 on Build GRCh38
rs759484452 Chr.12: 49637700-49637700 on Build GRCh38
rs776127111 Chr.12: 49637535-49637535 on Build GRCh38
rs746274382 Chr.12: 49637612-49637612 on Build GRCh38
rs200644715 Chr.12: 49637726-49637726 on Build GRCh38
rs1014404744 Chr.12: 49637721-49637721 on Build GRCh38
rs763219088 Chr.12: 49637614-49637614 on Build GRCh38
rs750255141 Chr.12: 49637565-49637565 on Build GRCh38
rs769234158 Chr.12: 49637618-49637618 on Build GRCh38
rs920244892 Chr.12: 49637664-49637664 on Build GRCh38
rs375291078 Chr.12: 49637715-49637715 on Build GRCh38
rs371206574 Chr.12: 49637582-49637582 on Build GRCh38
rs754869428 Chr.12: 49637598-49637598 on Build GRCh38
rs376216592 Chr.12: 49637656-49637656 on Build GRCh38
rs545375851 Chr.12: 49637634-49637634 on Build GRCh38
rs371192317 Chr.12: 49637600-49637600 on Build GRCh38
rs911606427 Chr.12: 49637646-49637646 on Build GRCh38
rs201358204 Chr.12: 49637714-49637714 on Build GRCh38
rs757345886 Chr.12: 49637704-49637704 on Build GRCh38
rs747664026 Chr.12: 49637724-49637724 on Build GRCh38
rs767137725 Chr.12: 49637558-49637558 on Build GRCh38
rs200247987 Chr.12: 49637645-49637645 on Build GRCh38
rs377670850 Chr.12: 49637608-49637608 on Build GRCh38
rs762600633 Chr.12: 49637544-49637544 on Build GRCh38
rs1034017226 Chr.12: 49637613-49637613 on Build GRCh38
rs756245523 Chr.12: 49637716-49637716 on Build GRCh38
rs374757667 Chr.12: 49637615-49637615 on Build GRCh38
rs1038712935 Chr.12: 49637661-49637661 on Build GRCh38
rs116701528 Chr.12: 49637654-49637654 on Build GRCh38
rs996492514 Chr.12: 49637730-49637730 on Build GRCh38
rs199787423 Chr.12: 49637703-49637703 on Build GRCh38
rs777852674 Chr.12: 49637599-49637599 on Build GRCh38
rs527447087 Chr.12: 49637717-49637717 on Build GRCh38
rs755544540 Chr.12: 49637719-49637719 on Build GRCh38
rs768245712 Chr.12: 49637553-49637553 on Build GRCh38
rs779552414 Chr.12: 49637720-49637720 on Build GRCh38
rs760430553 Chr.12: 49637685-49637685 on Build GRCh38
rs367831439 Chr.12: 49637620-49637620 on Build GRCh38
rs766741537 Chr.12: 49637713-49637716 on Build GRCh38
rs377520162 Chr.12: 49637590-49637590 on Build GRCh38
rs766280125 Chr.12: 49637583-49637583 on Build GRCh38
rs761390746 Chr.12: 49637557-49637557 on Build GRCh38
rs756478016 Chr.12: 49637614-49637614 on Build GRCh38
rs376128730 Chr.12: 49637555-49637555 on Build GRCh38
rs771583115 Chr.12: 49637737-49637737 on Build GRCh38
rs766197890 Chr.12: 49637697-49637697 on Build GRCh38
rs774476500 Chr.12: 49637728-49637729 on Build GRCh38
rs771682664 Chr.12: 49637627-49637627 on Build GRCh38
rs373466048 Chr.12: 49637629-49637629 on Build GRCh38
rs749839609 Chr.12: 49637617-49637617 on Build GRCh38
rs747626044 Chr.12: 49637624-49637624 on Build GRCh38
rs772560721 Chr.12: 49637743-49637743 on Build GRCh38
rs776362223 Chr.12: 49637699-49637699 on Build GRCh38

View Full Product Details