Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5745882-5746143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPERD
Entrez Gene ID
257062
Gene Name
cation channel sperm associated auxiliary subunit delta
Gene Aliases
TMEM146
UniGene
Hs.631842
Gene Chromosome Location
Chr.19: 5720385-5778734 on Build GRCh38
Transcripts
NM_152784.3 XM_011527885.2 XM_011527886.2 XM_011527887.2 XM_011527888.2 XM_011527889.2 XM_011527890.2 XM_017026564.1 XM_017026565.1 XM_017026566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
179257 c.699C>T p.F233F Chr.19: 5745954-5745954 on Build GRCh38
6152684 c.692G>T p.R231L Chr.19: 5745947-5745947 on Build GRCh38
5909687 c.709T>A p.F237I Chr.19: 5745964-5745964 on Build GRCh38
370484 c.713A>T p.D238V Chr.19: 5745968-5745968 on Build GRCh38
4081968 c.702G>T p.G234G Chr.19: 5745957-5745957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373573573 Chr.19: 5746096-5746096 on Build GRCh38
rs759549516 Chr.19: 5746038-5746038 on Build GRCh38
rs777047011 Chr.19: 5745919-5745919 on Build GRCh38
rs757001566 Chr.19: 5746075-5746075 on Build GRCh38
rs765495581 Chr.19: 5745937-5745937 on Build GRCh38
rs753615039 Chr.19: 5745966-5745966 on Build GRCh38
rs773094567 Chr.19: 5746043-5746045 on Build GRCh38
rs143692878 Chr.19: 5746120-5746120 on Build GRCh38
rs200681020 Chr.19: 5745968-5745968 on Build GRCh38
rs777630392 Chr.19: 5745960-5745960 on Build GRCh38
rs773518485 Chr.19: 5746016-5746016 on Build GRCh38
rs766577195 Chr.19: 5746026-5746026 on Build GRCh38
rs570335771 Chr.19: 5746002-5746002 on Build GRCh38
rs373521530 Chr.19: 5746033-5746033 on Build GRCh38
rs756076644 Chr.19: 5745957-5745957 on Build GRCh38
rs768054369 Chr.19: 5746110-5746110 on Build GRCh38
rs769080778 Chr.19: 5745917-5745917 on Build GRCh38
rs74336104 Chr.19: 5745996-5745996 on Build GRCh38
rs762200295 Chr.19: 5746003-5746003 on Build GRCh38
rs983802960 Chr.19: 5745956-5745956 on Build GRCh38
rs751648177 Chr.19: 5746037-5746037 on Build GRCh38
rs781695133 Chr.19: 5746112-5746112 on Build GRCh38
rs766461198 Chr.19: 5745954-5745954 on Build GRCh38
rs771757701 Chr.19: 5745983-5745983 on Build GRCh38
rs771944238 Chr.19: 5745998-5745998 on Build GRCh38
rs761218348 Chr.19: 5745914-5745914 on Build GRCh38
rs200180226 Chr.19: 5746013-5746013 on Build GRCh38
rs750255698 Chr.19: 5746093-5746093 on Build GRCh38
rs370051045 Chr.19: 5745991-5745991 on Build GRCh38
rs766377761 Chr.19: 5746109-5746110 on Build GRCh38
rs77656808 Chr.19: 5746110-5746111 on Build GRCh38
rs377544635 Chr.19: 5746045-5746045 on Build GRCh38
rs760768181 Chr.19: 5746052-5746052 on Build GRCh38
rs368212653 Chr.19: 5745942-5745942 on Build GRCh38
rs757052375 Chr.19: 5745967-5745967 on Build GRCh38
rs1051489708 Chr.19: 5746119-5746119 on Build GRCh38
rs763086834 Chr.19: 5745946-5745946 on Build GRCh38
rs745546371 Chr.19: 5745978-5745978 on Build GRCh38
rs753806882 Chr.19: 5746062-5746062 on Build GRCh38
rs960121712 Chr.19: 5745924-5745924 on Build GRCh38
rs762325095 Chr.19: 5745935-5745935 on Build GRCh38
rs369406616 Chr.19: 5746094-5746094 on Build GRCh38
rs201497391 Chr.19: 5745955-5745955 on Build GRCh38
rs778660807 Chr.19: 5746076-5746076 on Build GRCh38
rs746469165 Chr.19: 5746097-5746097 on Build GRCh38
rs374016093 Chr.19: 5745938-5745938 on Build GRCh38
rs368808350 Chr.19: 5746023-5746023 on Build GRCh38

View Full Product Details