Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 89601361-89601632 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2B1
Entrez Gene ID
490
Gene Name
ATPase plasma membrane Ca2+ transporting 1
Gene Aliases
PMCA1, PMCA1kb
UniGene
Hs.506276
Gene Chromosome Location
Chr.12: 89588049-89709353 on Build GRCh38
Transcripts
NM_001001323.1 NM_001682.2 XM_011538407.2 XM_017019357.1 XM_017019358.1 XM_017019359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3772742 c.3112A>G p.I1038V Chr.12: 89601382-89601382 on Build GRCh38
274091 c.3130T>C p.S1044P Chr.12: 89601364-89601364 on Build GRCh38
4947067 c.3087A>G p.K1029K Chr.12: 89601407-89601407 on Build GRCh38
3772744 c.3107T>G p.L1036R Chr.12: 89601387-89601387 on Build GRCh38
6286507 c.3075G>T p.Q1025H Chr.12: 89601419-89601419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186141948 Chr.12: 89601462-89601462 on Build GRCh38
rs778954138 Chr.12: 89601453-89601453 on Build GRCh38
rs974547112 Chr.12: 89601488-89601488 on Build GRCh38
rs748325409 Chr.12: 89601440-89601440 on Build GRCh38
rs866847354 Chr.12: 89601474-89601474 on Build GRCh38
rs552339430 Chr.12: 89601498-89601498 on Build GRCh38
rs868760313 Chr.12: 89601596-89601596 on Build GRCh38
rs917951528 Chr.12: 89601463-89601463 on Build GRCh38
rs746609465 Chr.12: 89601478-89601484 on Build GRCh38
rs745609831 Chr.12: 89601457-89601457 on Build GRCh38
rs886357662 Chr.12: 89601588-89601588 on Build GRCh38
rs1018899788 Chr.12: 89601417-89601417 on Build GRCh38
rs755053141 Chr.12: 89601392-89601392 on Build GRCh38
rs1047653297 Chr.12: 89601524-89601524 on Build GRCh38
rs757111302 Chr.12: 89601450-89601450 on Build GRCh38
rs941837503 Chr.12: 89601605-89601605 on Build GRCh38
rs781117632 Chr.12: 89601409-89601409 on Build GRCh38
rs964520391 Chr.12: 89601419-89601419 on Build GRCh38
rs1022558795 Chr.12: 89601489-89601489 on Build GRCh38
rs771882359 Chr.12: 89601467-89601467 on Build GRCh38
rs775054223 Chr.12: 89601479-89601479 on Build GRCh38
rs375545786 Chr.12: 89601536-89601536 on Build GRCh38
rs537486380 Chr.12: 89601582-89601582 on Build GRCh38

View Full Product Details