Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12079127-12079400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL2L14
Entrez Gene ID
79370
Gene Name
BCL2 like 14
Gene Aliases
BCLG
UniGene
Hs.210343
Gene Chromosome Location
Chr.12: 12049844-12099695 on Build GRCh38
Transcripts
NM_030766.1 NM_138722.1 NM_138723.1 XM_006719151.3 XM_011520848.1 XM_011520849.1 XM_017019963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7357394 c.65T>C p.I22T Chr.12: 12079370-12079370 on Build GRCh38
1299029 c.82G>A p.A28T Chr.12: 12079387-12079387 on Build GRCh38
3457225 c.37C>T p.P13S Chr.12: 12079342-12079342 on Build GRCh38
7357392 c.65T>C p.I22T Chr.12: 12079370-12079370 on Build GRCh38
259617 c.13A>G p.S5G Chr.12: 12079318-12079318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985570701 Chr.12: 12079274-12079274 on Build GRCh38
rs755725657 Chr.12: 12079251-12079251 on Build GRCh38
rs758294653 Chr.12: 12079280-12079280 on Build GRCh38
rs747924373 Chr.12: 12079279-12079279 on Build GRCh38
rs765970031 Chr.12: 12079259-12079259 on Build GRCh38
rs770002798 Chr.12: 12079307-12079307 on Build GRCh38
rs754606088 Chr.12: 12079343-12079343 on Build GRCh38
rs754676193 Chr.12: 12079265-12079265 on Build GRCh38
rs370415348 Chr.12: 12079351-12079351 on Build GRCh38
rs746093170 Chr.12: 12079306-12079306 on Build GRCh38
rs548677736 Chr.12: 12079159-12079159 on Build GRCh38
rs778583625 Chr.12: 12079275-12079275 on Build GRCh38
rs1023379555 Chr.12: 12079298-12079298 on Build GRCh38
rs755232118 Chr.12: 12079329-12079329 on Build GRCh38
rs753606618 Chr.12: 12079260-12079260 on Build GRCh38
rs766056839 Chr.12: 12079342-12079342 on Build GRCh38
rs762400796 Chr.12: 12079331-12079331 on Build GRCh38
rs777802246 Chr.12: 12079285-12079285 on Build GRCh38
rs771056389 Chr.12: 12079288-12079288 on Build GRCh38
rs971462263 Chr.12: 12079309-12079309 on Build GRCh38
rs776687121 Chr.12: 12079291-12079291 on Build GRCh38
rs765097567 Chr.12: 12079340-12079341 on Build GRCh38
rs61733244 Chr.12: 12079349-12079349 on Build GRCh38
rs566990379 Chr.12: 12079198-12079198 on Build GRCh38
rs59599134 Chr.12: 12079313-12079313 on Build GRCh38
rs746925895 Chr.12: 12079287-12079287 on Build GRCh38
rs1027175339 Chr.12: 12079227-12079227 on Build GRCh38
rs139406698 Chr.12: 12079323-12079323 on Build GRCh38
rs61733245 Chr.12: 12079370-12079370 on Build GRCh38
rs770635916 Chr.12: 12079228-12079229 on Build GRCh38
rs763337059 Chr.12: 12079319-12079319 on Build GRCh38

View Full Product Details