Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 12704841-12705074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPPED1
Entrez Gene ID
55313
Gene Name
calcineurin like phosphoesterase domain containing 1
Gene Aliases
CSTP1
UniGene
Hs.460002
Gene Chromosome Location
Chr.16: 12659799-12803887 on Build GRCh38
Transcripts
NM_001099455.1 NM_018340.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5850180 c.442G>A p.D148N Chr.16: 12704897-12704897 on Build GRCh38
3887919 c.480C>T p.F160F Chr.16: 12704859-12704859 on Build GRCh38
7362506 c.350G>T p.R117M Chr.16: 12704989-12704989 on Build GRCh38
5676469 c.431G>T p.R144L Chr.16: 12704908-12704908 on Build GRCh38
5769035 c.431G>A p.R144Q Chr.16: 12704908-12704908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766084274 Chr.16: 12704939-12704939 on Build GRCh38
rs774589162 Chr.16: 12704965-12704965 on Build GRCh38
rs143195532 Chr.16: 12704996-12704996 on Build GRCh38
rs761928107 Chr.16: 12705005-12705005 on Build GRCh38
rs145004672 Chr.16: 12704867-12704867 on Build GRCh38
rs779758297 Chr.16: 12704957-12704957 on Build GRCh38
rs776981458 Chr.16: 12705002-12705002 on Build GRCh38
rs747679788 Chr.16: 12704950-12704950 on Build GRCh38
rs199886762 Chr.16: 12705006-12705006 on Build GRCh38
rs758255991 Chr.16: 12704935-12704935 on Build GRCh38
rs572373767 Chr.16: 12704865-12704865 on Build GRCh38
rs764436302 Chr.16: 12704931-12704931 on Build GRCh38
rs758254494 Chr.16: 12704868-12704869 on Build GRCh38
rs754923212 Chr.16: 12704941-12704941 on Build GRCh38
rs772741061 Chr.16: 12704984-12704984 on Build GRCh38
rs375032569 Chr.16: 12705007-12705007 on Build GRCh38
rs755685139 Chr.16: 12705033-12705033 on Build GRCh38
rs1055853492 Chr.16: 12704999-12704999 on Build GRCh38
rs1001584731 Chr.16: 12705003-12705003 on Build GRCh38
rs371523609 Chr.16: 12705042-12705042 on Build GRCh38
rs759943831 Chr.16: 12704924-12704924 on Build GRCh38
rs748886359 Chr.16: 12704963-12704963 on Build GRCh38
rs904498691 Chr.16: 12704995-12704995 on Build GRCh38
rs565046403 Chr.16: 12704961-12704961 on Build GRCh38
rs750421490 Chr.16: 12704887-12704887 on Build GRCh38
rs202086201 Chr.16: 12705045-12705045 on Build GRCh38
rs368567080 Chr.16: 12705037-12705037 on Build GRCh38
rs372715528 Chr.16: 12705021-12705021 on Build GRCh38
rs3748982 Chr.16: 12704959-12704959 on Build GRCh38
rs915139484 Chr.16: 12704884-12704884 on Build GRCh38
rs1018532906 Chr.16: 12705044-12705044 on Build GRCh38
rs760889805 Chr.16: 12704991-12704991 on Build GRCh38
rs376756823 Chr.16: 12704878-12704878 on Build GRCh38
rs911663541 Chr.16: 12704922-12704922 on Build GRCh38
rs149102231 Chr.16: 12704921-12704921 on Build GRCh38
rs772801239 Chr.16: 12704906-12704906 on Build GRCh38
rs781067646 Chr.16: 12704943-12704943 on Build GRCh38
rs1001535667 Chr.16: 12704998-12704998 on Build GRCh38
rs746181084 Chr.16: 12704969-12704969 on Build GRCh38
rs370257644 Chr.16: 12704934-12704934 on Build GRCh38
rs182902514 Chr.16: 12705010-12705010 on Build GRCh38
rs781461264 Chr.16: 12704899-12704899 on Build GRCh38
rs753327791 Chr.16: 12705038-12705038 on Build GRCh38
rs564833035 Chr.16: 12704889-12704889 on Build GRCh38
rs945986292 Chr.16: 12704942-12704942 on Build GRCh38
rs529007917 Chr.16: 12704964-12704964 on Build GRCh38
rs962306773 Chr.16: 12705012-12705012 on Build GRCh38
rs186670586 Chr.16: 12705025-12705025 on Build GRCh38
rs369326763 Chr.16: 12705052-12705052 on Build GRCh38
rs369351725 Chr.16: 12704866-12704866 on Build GRCh38
rs754550316 Chr.16: 12704874-12704874 on Build GRCh38
rs747590104 Chr.16: 12704882-12704882 on Build GRCh38
rs771167301 Chr.16: 12704909-12704909 on Build GRCh38
rs768075830 Chr.16: 12704925-12704925 on Build GRCh38
rs202093661 Chr.16: 12704987-12704987 on Build GRCh38
rs759278011 Chr.16: 12705011-12705011 on Build GRCh38
rs370057241 Chr.16: 12704908-12704908 on Build GRCh38
rs746891361 Chr.16: 12704873-12704873 on Build GRCh38
rs780667074 Chr.16: 12704877-12704877 on Build GRCh38
rs977743533 Chr.16: 12705043-12705043 on Build GRCh38
rs201640855 Chr.16: 12704930-12704930 on Build GRCh38
rs775720619 Chr.16: 12704929-12704929 on Build GRCh38

View Full Product Details